This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03859854_s1
See other NACC2 GE Assays ›
Gene Symbol:
NACC2
Gene Name:
NACC family member 2
Gene Aliases:
BEND9, BTBD14, BTBD14A, BTBD31, NAC-2, RBB
Chromosome Location:
Chr.9: 136006537 - 136095285 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03859854_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NACC2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144653.4 NP_653254.1 6 3301 95
XM_011518221.2 XP_011516523.1 6 3321 95
GenBank mRNA AK094914.1 - 1 454 95
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA HY139632.1 - 1 387 Chr. 9 - 136010042 - 136010521 [-] on Build GRCh38 95
GenBankmRNA DA399580.1 - 1 304 Chr. 9 - 136009947 - 136010438 [-] on Build GRCh38 95
GenBankmRNA BF345463.1 - Chr. - - [-] on Build 96
GenBankmRNA DB476693.1 - 1 60 Chr. 9 - 136009700 - 136010193 [-] on Build GRCh38 95
GenBankmRNA DA410900.1 - 1 60 Chr. 9 - 136009643 - 136010194 [-] on Build GRCh38 95
GenBankmRNA BI029061.1 - 1 Chr. 9 - 136009868 - 136010304 [+] on Build GRCh38 95
GenBankmRNA BX474817.1 - 1 162 Chr. 9 - 136009717 - 136010296 [-] on Build GRCh38 95
GenBankmRNA DB235878.1 - 1 61 Chr. 9 - 136009638 - 136010195 [-] on Build GRCh38 95
GenBankmRNA DB470380.1 - 1 60 Chr. 9 - 136009768 - 136010193 [-] on Build GRCh38 95

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream UBA domain containing 1 UBAC1 GBDR1,
UBADC1
-45.7kb Sense Chr. 9 - 135932969 - 135961380 on NCBI Build GRCh38
Downstream uncharacterized LOC105376323 LOC105376323 97.2kb Sense Chr. 9 - 136107829 - 136109424 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900011962.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NACC2

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
positive regulation of cell proliferation
negative regulation of cell proliferation
posttranscriptional regulation of gene expression
histone deacetylation
cellular protein complex localization
negative regulation of transcription, DNA-templated
protein homooligomerization
negative regulation of G1/S transition of mitotic cell cycle by negative regulation of transcription from RNA polymerase II promoter
positive regulation of intrinsic apoptotic signaling pathway in response to DNA damage
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, RNA polymerase II transcription factor binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
histone deacetylase activity
protein binding
protein homodimerization activity
histone deacetylase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 ENST00000277554.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 ENST00000277554.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 ENST00000277554.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 ENST00000277554.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 ENST00000277554.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 ENST00000277554.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 ENST00000277554.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 ENST00000277554.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 ENST00000277554.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 ENST00000277554.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 ENST00000277554.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 ENST00000277554.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 ENST00000277554.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 ENST00000371753.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 ENST00000371753.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 ENST00000371753.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 ENST00000371753.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 ENST00000371753.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 ENST00000371753.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 ENST00000371753.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 ENST00000371753.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 ENST00000371753.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 ENST00000371753.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 ENST00000371753.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 ENST00000371753.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 ENST00000467669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 ENST00000467669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 NM_144653.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 NM_144653.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 NM_144653.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 NM_144653.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 NM_144653.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 NM_144653.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 NM_144653.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 NM_144653.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 NM_144653.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 NM_144653.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 NM_144653.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 NM_144653.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011962.hg.1 NM_144653.4 Refseq
Affymetrix Human Clariom D Assay PSR0900181791.hg.1 ENST00000277554.3 Ensembl
Affymetrix Human Clariom D Assay PSR0900181791.hg.1 ENST00000371753.5 Ensembl
Affymetrix Human Clariom D Assay PSR0900181791.hg.1 NM_144653.4 Refseq
Affymetrix Human Clariom D Assay PSR0900181791.hg.1 ENST00000277554.3 Ensembl
Affymetrix Human Clariom D Assay PSR0900181791.hg.1 ENST00000371753.5 Ensembl
Affymetrix Human Clariom D Assay PSR0900181791.hg.1 NM_144653.4 Refseq
Affymetrix Human Clariom D Assay PSR0900181791.hg.1 TC0900011962.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0900011962.hg.1 ENST00000277554.3 Ensembl
Affymetrix Human Clariom D Assay TC0900011962.hg.1 ENST00000277554.3 Ensembl
Affymetrix Human Clariom D Assay TC0900011962.hg.1 ENST00000277554.3 Ensembl
Affymetrix Human Clariom D Assay TC0900011962.hg.1 ENST00000277554.3 Ensembl
Affymetrix Human Clariom D Assay TC0900011962.hg.1 ENST00000277554.3 Ensembl
Affymetrix Human Clariom D Assay TC0900011962.hg.1 ENST00000277554.3 Ensembl
Affymetrix Human Clariom D Assay TC0900011962.hg.1 ENST00000277554.3 Ensembl
Affymetrix Human Clariom D Assay TC0900011962.hg.1 ENST00000277554.3 Ensembl
Affymetrix Human Clariom D Assay TC0900011962.hg.1 ENST00000277554.3 Ensembl
Affymetrix Human Clariom D Assay TC0900011962.hg.1 ENST00000277554.3 Ensembl
Affymetrix Human Clariom D Assay TC0900011962.hg.1 ENST00000277554.3 Ensembl
Affymetrix Human Clariom D Assay TC0900011962.hg.1 ENST00000277554.3 Ensembl
Affymetrix Human Clariom D Assay TC0900011962.hg.1 ENST00000277554.3 Ensembl
Affymetrix Human Clariom D Assay TC0900011962.hg.1 ENST00000371753.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011962.hg.1 ENST00000371753.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011962.hg.1 ENST00000371753.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011962.hg.1 ENST00000371753.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011962.hg.1 ENST00000371753.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011962.hg.1 ENST00000371753.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011962.hg.1 ENST00000371753.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011962.hg.1 ENST00000371753.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011962.hg.1 ENST00000371753.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011962.hg.1 ENST00000371753.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011962.hg.1 ENST00000371753.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011962.hg.1 ENST00000371753.5 Ensembl
Affymetrix Human Clariom D Assay TC0900011962.hg.1 ENST00000467669.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011962.hg.1 ENST00000467669.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011962.hg.1 NM_144653.4 Refseq
Affymetrix Human Clariom D Assay TC0900011962.hg.1 NM_144653.4 Refseq
Affymetrix Human Clariom D Assay TC0900011962.hg.1 NM_144653.4 Refseq
Affymetrix Human Clariom D Assay TC0900011962.hg.1 NM_144653.4 Refseq
Affymetrix Human Clariom D Assay TC0900011962.hg.1 NM_144653.4 Refseq
Affymetrix Human Clariom D Assay TC0900011962.hg.1 NM_144653.4 Refseq
Affymetrix Human Clariom D Assay TC0900011962.hg.1 NM_144653.4 Refseq
Affymetrix Human Clariom D Assay TC0900011962.hg.1 NM_144653.4 Refseq
Affymetrix Human Clariom D Assay TC0900011962.hg.1 NM_144653.4 Refseq
Affymetrix Human Clariom D Assay TC0900011962.hg.1 NM_144653.4 Refseq
Affymetrix Human Clariom D Assay TC0900011962.hg.1 NM_144653.4 Refseq
Affymetrix Human Clariom D Assay TC0900011962.hg.1 NM_144653.4 Refseq
Affymetrix Human Clariom D Assay TC0900011962.hg.1 NM_144653.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0