This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03853393_s1
See other APCDD1L-AS1 GE Assays ›
Gene Symbol:
APCDD1L-AS1
Gene Name:
APCDD1L antisense RNA 1 (head to head)
Gene Aliases:
-
Chromosome Location:
Chr.20: 58515379 - 58619892 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03853393_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: APCDD1L-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK090612.1 - 1 1150 116
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BU619904.1 - 1 Chr. 20 - 58525612 - 58526318 [-] on Build GRCh38 116

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream APC down-regulated 1 like APCDD1L -9.5kb Anti-sense Chr. 20 - 58459101 - 58514938 on NCBI Build GRCh38
Downstream uncharacterized LOC79160 LOC79160 107.7kb Sense Chr. 20 - 58634772 - 58635738 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0