Assay Details
Gene Symbol: PIPSL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_002319.2 | 1 | 3650 | 87 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | solute carrier family 35 member G1 | SLC35G1 |
C10orf60, TMEM20 |
-43.9kb | Anti-sense | Chr. 10 - 93893951 - 93914265 on NCBI Build GRCh38 |
Downstream | phospholipase C epsilon 1 | PLCE1 |
NPHS3, PLCE, PPLC |
32.1kb | Anti-sense | Chr. 10 - 93993989 - 94329447 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10001539.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 389538 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 148 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3751 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3750 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P435367 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07401923 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07401911 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27791559 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16373693 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27791564 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07401916 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07401914 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07401926 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27791533 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07401925 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27791547 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27791545 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16373702 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27791558 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27791535 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27791568 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27791571 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27791557 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27791569 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27791565 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07401915 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27791547 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07401914 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27791557 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07401925 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07401923 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27791558 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27791569 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16373702 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27791533 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07401926 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27791571 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27791545 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27791535 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16373693 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27791564 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07401916 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07401915 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07401911 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27791568 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27791559 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27791565 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07401879 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07401895 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16373668 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07401892 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16373703 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07401887 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07401926 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16373704 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07401924 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07401889 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07401914 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07401898 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07401900 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07401869 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07401902 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07401876 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07401908 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07401911 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07401872 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07401884 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07401909 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07401867 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07401882 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07401931 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07401877 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07401906 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07401921 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07401875 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07401916 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07401918 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NR_002319_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 7935140 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7935141 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7935139 | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR10019376.hg.1 | NR_002319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10019376.hg.1 | TC10001539.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000011460.hg.1 | ENST00000480546.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011460.hg.1 | ENST00000480546.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011460.hg.1 | ENST00000489875.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011460.hg.1 | NR_002319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011460.hg.1 | NR_002319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011460.hg.1 | NR_002319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011460.hg.1 | NR_002319.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000177798.hg.1 | NR_002319.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000177798.hg.1 | NR_002319.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000177798.hg.1 | TC1000011460.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1000011460.hg.1 | ENST00000480546.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011460.hg.1 | ENST00000480546.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011460.hg.1 | ENST00000489875.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011460.hg.1 | NR_002319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011460.hg.1 | NR_002319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011460.hg.1 | NR_002319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011460.hg.1 | NR_002319.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000177799.hg.1 | ENST00000480546.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177799.hg.1 | NR_002319.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000177799.hg.1 | ENST00000480546.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177799.hg.1 | NR_002319.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000177799.hg.1 | TC1000011460.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A043126 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A043125 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A043125 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A043126 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 174829 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1676763 | NR_002319.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1676763 | NR_002319.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1676763 | NR_002319.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1676763 | NR_002319.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1676763 | NR_002319.2 | human_transcriptome |
