This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03677213_s1
See other RPS15AP10 GE Assays ›
Gene Symbol:
RPS15AP10
Gene Name:
ribosomal protein S15a pseudogene 10
Gene Aliases:
RPS15A_4_42
Chromosome Location:
Chr.1: 45645772 - 45646685 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03677213_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPS15AP10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_026768.1 1 455 98
GenBank mRNA CA843872.1 - 1 98
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BG654688.1 - 1 Chr. 1 - 45645783 - 45646311 [+] on Build GRCh38 98
GenBankmRNA BX481330.1 - 1 459 Chr. 1 - 45646020 - 45646688 [-] on Build GRCh38 98
GenBankmRNA CB162364.1 - 1 223 Chr. 1 - 45645857 - 45646443 [-] on Build GRCh38 98
GenBankmRNA AJ705415.1 - 1 325 Chr. 1 - 45646056 - 45646534 [-] on Build GRCh38 98
GenBankmRNA BG654885.1 - 1 148 Chr. 1 - 45645815 - 45646374 [-] on Build GRCh38 98
GenBankmRNA CA843117.1 - 1 147 Chr. 1 - 45645772 - 45646374 [-] on Build GRCh38 98

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream coiled-coil domain containing 17 CCDC17 -21.0kb Sense Chr. 1 - 45620044 - 45624787 on NCBI Build GRCh38
Downstream transmembrane protein 69 TMEM69 C1orf154 41.5kb Anti-sense Chr. 1 - 45688175 - 45694436 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100014026.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01002614.hg.1

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ArrayStar

Array Name Probe IDs Targets Target Type
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3357049 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4765 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4764 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 10221 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3357049 - -
SurePrint G3 Human GE Microarray G4851A A_33_P3357049 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20201306 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20201301 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15056186 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00175962 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00175938 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00175931 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20201308 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20201275 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20201321 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15056162 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20201291 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20201311 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20201289 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20201303 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15056183 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00175953 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20201295 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15056164 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00175940 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20201310 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20201307 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00175945 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15056169 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00175936 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15056161 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15056172 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00175942 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20201313 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20201288 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20201316 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00175944 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00175940 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15056183 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00175953 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15056161 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20201308 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20201303 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15056169 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20201275 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00175936 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20201291 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20201288 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15056162 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00175931 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00175942 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20201289 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20201306 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20201313 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20201316 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20201321 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15056172 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15056186 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00175938 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20201311 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20201310 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00175962 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00175945 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20201295 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20201307 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00175944 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15056164 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20201301 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00175950 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00175948 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00175955 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P15056164 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P15056184 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P15056185 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00175949 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00175954 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00175931 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00175933 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00175958 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00175956 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00175938 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00175967 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00175942 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00175963 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00175940 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P15056180 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00175966 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00175945 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00175936 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P15056188 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00175953 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P15056182 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01040752.hg.1 NR_026768.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01040752.hg.1 TC01002614.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0100014026.hg.1 ENST00000432472.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014026.hg.1 NR_026768.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014026.hg.1 NR_026768.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014026.hg.1 NR_026768.1 Refseq
Affymetrix Human Clariom D Assay TC0100014026.hg.1 ENST00000432472.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014026.hg.1 NR_026768.1 Refseq
Affymetrix Human Clariom D Assay TC0100014026.hg.1 NR_026768.1 Refseq
Affymetrix Human Clariom D Assay TC0100014026.hg.1 NR_026768.1 Refseq
Affymetrix Human Clariom D Assay PSR0100204917.hg.1 ENST00000432472.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100204917.hg.1 NR_026768.1 Refseq
Affymetrix Human Clariom D Assay PSR0100204917.hg.1 ENST00000432472.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100204917.hg.1 NR_026768.1 Refseq
Affymetrix Human Clariom D Assay PSR0100204917.hg.1 TC0100014026.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100204916.hg.1 NR_026768.1 Refseq
Affymetrix Human Clariom D Assay PSR0100204916.hg.1 NR_026768.1 Refseq
Affymetrix Human Clariom D Assay PSR0100204916.hg.1 TC0100014026.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100204918.hg.1 NR_026768.1 Refseq
Affymetrix Human Clariom D Assay PSR0100204918.hg.1 NR_026768.1 Refseq
Affymetrix Human Clariom D Assay PSR0100204918.hg.1 TC0100014026.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
Arraystar Human LncRNA microarray V2.0 ASHG19A3A044957 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A044957 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0