Assay Details
Gene Symbol: GJD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC035898.1 | - | 1 | 1453 | 101 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cyclin Y | CCNY |
C10orf9, CBCP1, CCNX, CFP1 |
-33.3kb | Anti-sense | Chr. 10 - 35246981 - 35572670 on NCBI Build GRCh38 |
Downstream | frizzled class receptor 8 | FZD8 |
FZ-8, hFZ8 |
30.1kb | Sense | Chr. 10 - 35638249 - 35641434 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000010325.hg.1 | ENST00000609313.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010325.hg.1 | ENST00000609313.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010325.hg.1 | ENST00000609313.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000558025.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000558025.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559702.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559702.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559702.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040055.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040055.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040055.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040056.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040056.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040056.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010325.hg.1 | ENST00000609313.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010325.hg.1 | ENST00000609313.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010325.hg.1 | ENST00000609313.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000558025.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000558025.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559702.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559702.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559702.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040055.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040055.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040055.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040056.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040056.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040056.1 | Refseq |
