This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03673702_s1
See other PODN GE Assays ›
Gene Symbol:
PODN
Gene Name:
podocan
Gene Aliases:
PCAN, SLRR5A
Chromosome Location:
Chr.1: 53062052 - 53085502 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03673702_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PODN
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AL122104.1 - 1 352 76

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream sterol carrier protein 2 SCP2 NLTP,
NSL-TP,
SCP-2,
SCP-CHI,
SCP-X,
SCPX
-19.3kb Sense Chr. 1 - 52927229 - 53051617 on NCBI Build GRCh38
Downstream solute carrier family 1 member 7 SLC1A7 AAAT,
EAAT5
15.6kb Anti-sense Chr. 1 - 53087179 - 53142632 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000312553.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000371500.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000395871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000395871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000395871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000395871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000395871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000395871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000395871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000395871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000395871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000395871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000395871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000395871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000395871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000395871.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000471210.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000471210.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000471210.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000471285.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000471285.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000471285.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000471285.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000471285.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000471285.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000490650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000490650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000490650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000490650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000490650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 ENST00000618387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199080.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199081.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199082.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199082.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199082.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199082.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199082.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199082.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199082.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199082.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199082.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199082.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199082.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199082.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199082.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_001199082.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008341.hg.1 NM_153703.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014220.hg.1 ENST00000438361.2 Ensembl
Affymetrix Human Clariom D Assay TC0100014220.hg.1 ENST00000438361.2 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0