This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03672385_s1
See other SORCS3-AS1 GE Assays ›
Gene Symbol:
SORCS3-AS1
Gene Name:
SORCS3 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.10: 104664608 - 104666216 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03672385_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SORCS3-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK097854.1 - 1 743 111

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC101927523 LOC101927523 -184.9kb Sense Chr. 10 - 104474939 - 104480275 on NCBI Build GRCh38
Downstream uncharacterized LOC101927549 LOC101927549 1.0mb Sense Chr. 10 - 105673610 - 105820333 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3805819 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000011755.hg.1 ENST00000449805.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011755.hg.1 ENST00000449805.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011755.hg.1 NR_104028.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011755.hg.1 NR_104028.1 Refseq
Affymetrix Human Clariom D Assay TC1000011755.hg.1 ENST00000449805.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011755.hg.1 ENST00000449805.2 Ensembl
Affymetrix Human Clariom D Assay TC1000011755.hg.1 NR_104028.1 Refseq
Affymetrix Human Clariom D Assay TC1000011755.hg.1 NR_104028.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0