This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03671765_s1
See other TMEM185B GE Assays ›
Gene Symbol:
TMEM185B
Gene Name:
transmembrane protein 185B
Gene Aliases:
FAM11B
Chromosome Location:
Chr.2: 120217471 - 120223408 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03671765_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMEM185B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024121.2 NP_077026.2 1 3716 121
GenBank mRNA AK021716.1 - 1 450 121
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA577162.1 - 1 368 Chr. 2 - 120219496 - 120220060 [-] on Build GRCh38 121
GenBankmRNA BI257465.1 - 1 166 Chr. 2 - 120219167 - 120219858 [-] on Build GRCh38 121
GenBankmRNA AL119878.1 - 1 229 Chr. 2 - 120219411 - 120219921 [-] on Build GRCh38 121
GenBankmRNA DA435851.1 - 1 266 Chr. 2 - 120219391 - 120219956 [-] on Build GRCh38 121
GenBankmRNA DA502959.1 - 1 267 Chr. 2 - 120219440 - 120219959 [-] on Build GRCh38 121
GenBankmRNA DA446170.1 - 1 463 Chr. 2 - 120219596 - 120220153 [-] on Build GRCh38 121

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream erythrocyte membrane protein band 4.1 like 5 EPB41L5 BE37,
YMO1,
YRT
-40.1kb Anti-sense Chr. 2 - 120013028 - 120179121 on NCBI Build GRCh38
Downstream RALB Ras like proto-oncogene B RALB 19.9kb Anti-sense Chr. 2 - 120240070 - 120294711 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200014049.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR0200202846.hg.1 NM_024121.2 Refseq
Affymetrix Human Clariom D Assay PSR0200202846.hg.1 NM_024121.2 Refseq
Affymetrix Human Clariom D Assay PSR0200202846.hg.1 TC0200014049.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0