This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03671611_s1
See other FUNDC2 GE Assays ›
Gene Symbol:
FUNDC2
Gene Name:
FUN14 domain containing 2
Gene Aliases:
DC44, HCBP6, HCC3, PD03104
Chromosome Location:
Chr.X: 155026789 - 155056916 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03671611_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FUNDC2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AF267862.1 - 1 50 125
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AA258633.1 - 1 50 Chr. 5 - 73508509 - 73508886 [+] on Build GRCh38 125

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream basic transcription factor 3 BTF3 BETA-NAC,
BTF3a,
BTF3b,
NACB
-2.9kb Sense Chr. 5 - 73498425 - 73505623 on NCBI Build GRCh38
Downstream ankyrin repeat family A member 2 ANKRA2 ANKRA 42.7kb Anti-sense Chr. 5 - 73552200 - 73565705 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF267862.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0