This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03670459_s1
See other TNK2-AS1 GE Assays ›
Gene Symbol:
TNK2-AS1
Gene Name:
TNK2 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
-
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03670459_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TNK2-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK094115.1 - 1 850 66

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream tyrosine kinase non receptor 2 TNK2 ACK,
ACK-1,
ACK1,
p21cdc42Hs
-2.3kb Anti-sense Chr. 3 - 195863364 - 195909009 on NCBI Build GRCh38
Downstream succinate dehydrogenase complex flavoprotein subunit A pseudogene 1 SDHAP1 SDHAL1,
SDHALP1
46.7kb Anti-sense Chr. 3 - 195959921 - 195990279 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Human Whole Genome ChIP Tiling Array A_17_P02877690 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P23019519 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P23019524 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P02877692 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3404686 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P110094 - -
Agilent Whole Human Genome Oligo Microarray 4x44K 4275 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 18313 - -
SurePrint G3 Human GE Microarray G4851A A_33_P3404686 - -
Whole Human Genome Oligo Microarray G4112A A_24_P110094 - -
Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P110094 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P36607805 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.0 ST Array 8084922 - -
Affymetrix Human Gene 1.1 ST Array 8084921 - -
Affymetrix HT HG-U133+ PM Array Plate 1565784_PM_at - -
Affymetrix Human Transcriptome Array 2.0 TC0300009985.hg.1 ENST00000424819.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009985.hg.1 ENST00000424819.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009985.hg.1 ENST00000448113.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009985.hg.1 ENST00000448113.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009985.hg.1 ENST00000448113.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009985.hg.1 ENST00000424819.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009985.hg.1 ENST00000424819.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009985.hg.1 ENST00000448113.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009985.hg.1 ENST00000448113.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009985.hg.1 ENST00000448113.1 Ensembl
Human Genome U133 Plus 2.0 Array 1565784_at - -
Human Genome U133 Plus 2.0 Array 1565784_PM_at - -
Affymetrix Human Genome U133 Plus 2.0 Array 1565784_at3 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1565784_at2 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1565784_at7 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1565784_at8 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1565784_at9 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1565784_at4 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1565784_at6 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1565784_at5 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1565784_at11 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1565784_at1 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1565784_at10 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0