This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03670189_s1
See other TIMM50 GE Assays ›
Gene Symbol:
TIMM50
Gene Name:
translocase of inner mitochondrial membrane 50
Gene Aliases:
TIM50, TIM50L
Chromosome Location:
Chr.19: 39480412 - 39493019 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03670189_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TIMM50
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK092396.1 - 1 651 82
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BU508684.1 - 259 Chr. - - [-] on Build 82
GenBankmRNA CB145535.1 - 1 80 Chr. 19 - 39492903 - 39493342 [+] on Build GRCh38 82

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream SPT5 homolog,
DSIF elongation factor subunit
SUPT5H SPT5,
SPT5H,
Tat-CT1
-15.7kb Sense Chr. 19 - 39445546 - 39476670 on NCBI Build GRCh38
Downstream delta like canonical Notch ligand 3 DLL3 SCDO1 5.1kb Sense Chr. 19 - 39498917 - 39508481 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1900011730.hg.1 ENST00000595527.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011730.hg.1 ENST00000595527.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011730.hg.1 ENST00000595527.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011730.hg.1 ENST00000595527.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011730.hg.1 ENST00000595527.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011730.hg.1 ENST00000600878.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011730.hg.1 ENST00000600878.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011730.hg.1 ENST00000595527.2 Ensembl
Affymetrix Human Clariom D Assay TC1900011730.hg.1 ENST00000595527.2 Ensembl
Affymetrix Human Clariom D Assay TC1900011730.hg.1 ENST00000595527.2 Ensembl
Affymetrix Human Clariom D Assay TC1900011730.hg.1 ENST00000595527.2 Ensembl
Affymetrix Human Clariom D Assay TC1900011730.hg.1 ENST00000595527.2 Ensembl
Affymetrix Human Clariom D Assay TC1900011730.hg.1 ENST00000600878.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011730.hg.1 ENST00000600878.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0