This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03669465_s1
See other SH3TC1 GE Assays ›
Gene Symbol:
SH3TC1
Gene Name:
SH3 domain and tetratricopeptide repeats 1
Gene Aliases:
-
Chromosome Location:
Chr.4: 8182072 - 8241103 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03669465_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SH3TC1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK057561.1 - 1 246 95
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB100951.1 - 1 246 Chr. 4 - 8208564 - 8209322 [+] on Build GRCh38 95

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream actin binding LIM protein family member 2 ABLIM2 -49.7kb Anti-sense Chr. 4 - 7965310 - 8158832 on NCBI Build GRCh38
Downstream HtrA serine peptidase 3 HTRA3 Prsp,
Tasp
59.0kb Sense Chr. 4 - 8269757 - 8307111 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0