This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03668714_s1
See other PLCB4 GE Assays ›
Gene Symbol:
PLCB4
Gene Name:
phospholipase C beta 4
Gene Aliases:
ARCND2, PI-PLC
Chromosome Location:
Chr.20: 9068710 - 9480816 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03668714_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PLCB4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK054754.1 - 1 152 82
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA055327.1 - 1 152 Chr. 20 - 9477299 - 9477859 [+] on Build GRCh38 82

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream phospholipase C beta 1 PLCB1 EIEE12,
PI-PLC,
PLC-154,
PLC-I,
PLC-beta-1,
PLC154,
PLCB1A,
PLCB1B
-592.4kb Sense Chr. 20 - 8132236 - 8884911 on NCBI Build GRCh38
Downstream LAMP5 antisense RNA 1 LAMP5-AS1 25.9kb Anti-sense Chr. 20 - 9505180 - 9514998 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7bff9b55f5-ncxmj:80/100.66.128.142:80.
git-commit: 5f36da1a8ad9bc9722227bfd86ebc827ae27d083
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0