This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03667715_s1
See other NUP54 GE Assays ›
Gene Symbol:
NUP54
Gene Name:
nucleoporin 54
Gene Aliases:
-
Chromosome Location:
Chr.4: 76114659 - 76148515 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03667715_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NUP54
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK022106.1 - 1 2342 82
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CD656030.1 - 1 Chr. 4 - 76140286 - 76141011 [+] on Build GRCh38 82
GenBankmRNA CD654756.1 - 1 Chr. 4 - 76140286 - 76141011 [+] on Build GRCh38 82

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ADP-ribosyltransferase 3 ART3 ARTC3 -27.5kb Anti-sense Chr. 4 - 76011180 - 76112802 on NCBI Build GRCh38
Downstream scavenger receptor class B member 2 SCARB2 AMRF,
CD36L2,
EPM4,
HLGP85,
LGP85,
LIMP-2,
LIMPII,
SR-BII
15.5kb Sense Chr. 4 - 76158737 - 76213899 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0