This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03667537_s1
See other FCF1 GE Assays ›
Gene Symbol:
FCF1
Gene Name:
FCF1 rRNA-processing protein
Gene Aliases:
Bka, C14orf111, CGI-35, UTP24
Chromosome Location:
Chr.14: 74713118 - 74738620 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03667537_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FCF1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK021636.1 - 1 652 90
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AU118329.1 - 1 652 Chr. 14 - 74713176 - 74713978 [+] on Build GRCh38 90

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream apoptosis resistant E3 ubiquitin protein ligase 1 AREL1 FIEL1,
KIAA0317
-61.0 Anti-sense Chr. 14 - 74661252 - 74713118 on NCBI Build GRCh38
Downstream YLP motif containing 1 YLPM1 C14orf170,
PPP1R169,
ZAP113,
ZAP3
48.5kb Sense Chr. 14 - 74763322 - 74837310 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000534938.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000534938.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000534938.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000534938.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000534938.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000534938.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000534938.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000534938.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000534938.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000534938.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000553615.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000553615.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000553615.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000553615.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000553615.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000553615.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000553615.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000553615.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000553615.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000553673.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000553673.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000553673.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000553673.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000553673.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000553673.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000553673.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000554064.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000554064.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000554064.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000554064.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000554064.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000554064.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000554064.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000554590.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000554590.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000554590.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000554590.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000554590.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000554590.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000554590.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000554973.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000554973.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000554973.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000554973.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000554973.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000554973.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000556814.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000556814.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000556814.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000556814.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000556814.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000556814.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 ENST00000556814.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 NM_015962.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 NM_015962.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 NM_015962.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 NM_015962.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 NM_015962.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 NM_015962.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 NM_015962.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 NM_015962.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 NM_015962.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 NM_015962.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 NM_015962.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 NM_015962.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 NM_015962.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007688.hg.1 NM_015962.4 Refseq
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000341162.8 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000534938.6 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000534938.6 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000534938.6 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000534938.6 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000534938.6 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000534938.6 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000534938.6 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000534938.6 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000534938.6 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000534938.6 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000553615.5 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000553615.5 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000553615.5 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000553615.5 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000553615.5 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000553615.5 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000553615.5 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000553615.5 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000553615.5 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000553673.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000553673.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000553673.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000553673.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000553673.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000553673.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000553673.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000554064.5 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000554064.5 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000554064.5 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000554064.5 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000554064.5 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000554064.5 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000554064.5 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000554590.5 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000554590.5 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000554590.5 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000554590.5 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000554590.5 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000554590.5 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000554590.5 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000554973.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000554973.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000554973.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000554973.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000554973.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000554973.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000556814.5 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000556814.5 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000556814.5 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000556814.5 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000556814.5 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000556814.5 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 ENST00000556814.5 Ensembl
Affymetrix Human Clariom D Assay TC1400007688.hg.1 NM_015962.4 Refseq
Affymetrix Human Clariom D Assay TC1400007688.hg.1 NM_015962.4 Refseq
Affymetrix Human Clariom D Assay TC1400007688.hg.1 NM_015962.4 Refseq
Affymetrix Human Clariom D Assay TC1400007688.hg.1 NM_015962.4 Refseq
Affymetrix Human Clariom D Assay TC1400007688.hg.1 NM_015962.4 Refseq
Affymetrix Human Clariom D Assay TC1400007688.hg.1 NM_015962.4 Refseq
Affymetrix Human Clariom D Assay TC1400007688.hg.1 NM_015962.4 Refseq
Affymetrix Human Clariom D Assay TC1400007688.hg.1 NM_015962.4 Refseq
Affymetrix Human Clariom D Assay TC1400007688.hg.1 NM_015962.4 Refseq
Affymetrix Human Clariom D Assay TC1400007688.hg.1 NM_015962.4 Refseq
Affymetrix Human Clariom D Assay TC1400007688.hg.1 NM_015962.4 Refseq
Affymetrix Human Clariom D Assay TC1400007688.hg.1 NM_015962.4 Refseq
Affymetrix Human Clariom D Assay TC1400007688.hg.1 NM_015962.4 Refseq
Affymetrix Human Clariom D Assay TC1400007688.hg.1 NM_015962.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0