This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03665032_s1
See other LOC105377021 GE Assays ›
Gene Symbol:
LOC105377021
Gene Name:
uncharacterized LOC105377021
Gene Aliases:
-
Chromosome Location:
Chr.3: 32815065 - 32817290 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03665032_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC105377021
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011534334.2 XP_011532636.1 1 475 88
GenBank mRNA AY358209.1 - 1 551 88

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream CCR4-NOT transcription complex subunit 10 CNOT10 -42.5kb Anti-sense Chr. 3 - 32685145 - 32773875 on NCBI Build GRCh38
Downstream tripartite motif containing 71 TRIM71 LIN-41,
LIN41
652.0 Anti-sense Chr. 3 - 32818018 - 32897826 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Human Whole Genome ChIP Tiling Array A_17_P22289655 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P932875 - -
Agilent Whole Human Genome Oligo Microarray 4x44K 16837 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 40306 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_24_P932875 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.0 ST Array 8086027 - -
Affymetrix Human Gene 1.0 ST Array 8086025 - -
Affymetrix Human Gene 1.0 ST Array 8086026 - -
Affymetrix Human Gene 1.1 ST Array 8086024 - -
Array Name Probe IDs Targets Target Type
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1702727 AY358209.1 human_transcriptome
Illumina human-6 v2.0 expression beadchip ILMN_1702727 AY358209.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1702727 AY358209.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header