This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03662035_s1
See other LOC100129275 GE Assays ›
Gene Symbol:
LOC100129275
Gene Name:
uncharacterized LOC100129275
Gene Aliases:
-
Chromosome Location:
-
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03662035_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100129275
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK126260.1 - 1 553 85

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream nuclear receptor subfamily 1 group I member 2 NR1I2 BXR,
ONR1,
PAR,
PAR1,
PAR2,
PARq,
PRR,
PXR,
SAR,
SXR
-15.4kb Anti-sense Chr. 3 - 119780484 - 119818485 on NCBI Build GRCh38
Downstream G protein-coupled receptor 156 GPR156 GABABL,
PGR28
330.0kb Sense Chr. 3 - 120165481 - 120285122 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent

Array Name Probe IDs Targets Target Type
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3423590 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 30818 - -
SurePrint G3 Human GE Microarray G4851A A_33_P3423590 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0