This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03661010_s1
See other MORC3 GE Assays ›
Gene Symbol:
MORC3
Gene Name:
MORC family CW-type zinc finger 3
Gene Aliases:
NXP2, ZCW5, ZCWCC3
Chromosome Location:
Chr.21: 36320189 - 36376646 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03661010_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MORC3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK125011.1 - 1 2153 79

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream dopey family member 2 DOPEY2 21orf5,
C21orf5
-64.8kb Sense Chr. 21 - 36156781 - 36294274 on NCBI Build GRCh38
Downstream chromatin assembly factor 1 subunit B CHAF1B CAF-1,
CAF-IP60,
CAF1,
CAF1A,
CAF1P60,
MPHOSPH7,
MPP7
15.5kb Sense Chr. 21 - 36380145 - 36416827 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800008094.hg.1 ENST00000534675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008094.hg.1 ENST00000534675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000397184.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000397184.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000397184.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000484028.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000484028.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000484028.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000484028.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000484028.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000485299.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000485299.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000485299.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000485299.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000485933.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000485933.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000485933.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000492336.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000492336.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000492336.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000492336.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000492336.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000492336.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000492336.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 ENST00000492336.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Clariom D Assay TC0800008094.hg.1 ENST00000534675.1 Ensembl
Affymetrix Human Clariom D Assay TC0800008094.hg.1 ENST00000534675.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000397184.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000397184.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000397184.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000400485.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000484028.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000484028.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000484028.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000484028.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000484028.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000485299.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000485299.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000485299.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000485299.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000485933.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000485933.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000485933.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000487909.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000492336.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000492336.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000492336.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000492336.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000492336.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000492336.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000492336.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 ENST00000492336.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Clariom D Assay TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Clariom D Assay TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Clariom D Assay TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Clariom D Assay TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Clariom D Assay TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Clariom D Assay TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Clariom D Assay TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Clariom D Assay TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Clariom D Assay TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Clariom D Assay TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Clariom D Assay TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Clariom D Assay TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Clariom D Assay TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Clariom D Assay TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Clariom D Assay TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Clariom D Assay TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Clariom D Assay TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Clariom D Assay TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Clariom D Assay TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Clariom D Assay TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Clariom D Assay TC2100008506.hg.1 NM_015358.2 Refseq
Affymetrix Human Clariom D Assay TC2100008506.hg.1 NM_015358.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0