This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03656268_s1
See other NUDT3 GE Assays ›
Gene Symbol:
NUDT3
Gene Name:
nudix hydrolase 3
Gene Aliases:
DIPP, DIPP-1, DIPP1
Chromosome Location:
Chr.6: 34287194 - 34392680 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03656268_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NUDT3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC024593.1 - 1 3747 79

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC102723521 LOC102723521 -97.9kb Anti-sense Chr. 6 - 34273202 - 34282446 on NCBI Build GRCh38
Downstream ribosomal protein S10 RPS10 DBA9,
S10
33.0kb Sense Chr. 6 - 34417454 - 34426125 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700006537.hg.1 ENST00000571815.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700006537.hg.1 ENST00000571815.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700006537.hg.1 ENST00000571815.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700006537.hg.1 ENST00000571815.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700006537.hg.1 ENST00000623952.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012035.hg.1 ENST00000551531.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012035.hg.1 ENST00000551531.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012035.hg.1 ENST00000551531.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012035.hg.1 ENST00000551531.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012035.hg.1 ENST00000551940.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012035.hg.1 ENST00000551940.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012035.hg.1 ENST00000551940.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012035.hg.1 ENST00000551940.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012035.hg.1 ENST00000551940.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012035.hg.1 ENST00000551940.1 Ensembl
Affymetrix Human Clariom D Assay TC1700006537.hg.1 ENST00000571815.1 Ensembl
Affymetrix Human Clariom D Assay TC1700006537.hg.1 ENST00000571815.1 Ensembl
Affymetrix Human Clariom D Assay TC1700006537.hg.1 ENST00000571815.1 Ensembl
Affymetrix Human Clariom D Assay TC1700006537.hg.1 ENST00000571815.1 Ensembl
Affymetrix Human Clariom D Assay TC1700006537.hg.1 ENST00000623952.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012035.hg.1 ENST00000551531.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012035.hg.1 ENST00000551531.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012035.hg.1 ENST00000551531.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012035.hg.1 ENST00000551531.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012035.hg.1 ENST00000551940.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012035.hg.1 ENST00000551940.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012035.hg.1 ENST00000551940.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012035.hg.1 ENST00000551940.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012035.hg.1 ENST00000551940.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012035.hg.1 ENST00000551940.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0