This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03655703_s1
See other LOC100131860 GE Assays ›
Gene Symbol:
LOC100131860
Gene Name:
uncharacterized LOC100131860
Gene Aliases:
-
Chromosome Location:
-
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03655703_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100131860
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK097109.1 - 1 744 98
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AW473610.1 - 1 Chr. 15 - 82985241 - 82985700 [+] on Build GRCh38 98
GenBankmRNA DN998893.1 - 1 542 Chr. 15 - 82985173 - 82985906 [-] on Build GRCh38 98

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream WAS protein homolog associated with actin,
golgi membranes and microtubules
WHAMM WHAMM1,
WHDC1
-149.2kb Anti-sense Chr. 15 - 82808177 - 82834861 on NCBI Build GRCh38
Downstream family with sequence similarity 103 member A1 FAM103A1 C15orf18,
HsT19360,
RAM
89.0 Anti-sense Chr. 15 - 82986203 - 82991057 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3409620 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P187871 - -
Agilent Whole Human Genome Oligo Microarray 4x44K 6125 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 57336 - -
SurePrint G3 Human GE Microarray G4851A A_33_P3409620 - -
Whole Human Genome Oligo Microarray G4112A A_24_P187871 - -
Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P187871 - -
Agilent Human 1B Microarray G4111A A_32_P362983 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.0 ST Array XM_001720708_at - -
Affymetrix Human Gene 1.0 ST Array XM_001720646_at - -
Affymetrix Human Gene 1.0 ST Array XM_001716100_at - -
Affymetrix Human Gene 1.0 ST Array 7991048 - -
Affymetrix Human Gene 1.1 ST Array 7991047 - -
Affymetrix HT HG-U133+ PM Array Plate 1559180_PM_at - -
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000304231.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000304231.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000304231.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000304231.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000304231.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000304231.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000304231.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000304231.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000304231.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000304231.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000304231.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000304231.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000304231.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000304231.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000304231.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000450735.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000450735.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000450735.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000450735.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000450735.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000450735.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000450735.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000450735.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000450735.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000450735.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000450735.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000450735.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000450735.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000450735.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000500334.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000500334.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000500334.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000558090.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000558090.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000558090.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000558090.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000558552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000558552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000558552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000558552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000558552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000558552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000558817.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000558817.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000558817.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000558817.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000558817.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000558817.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000558817.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000558817.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000560374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000560374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000560374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000560374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000560374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000560374.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000561345.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000561345.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000561345.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000561345.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000561345.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000561345.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000561345.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000619240.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000619240.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000619240.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000619367.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000619367.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 ENST00000619367.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 NM_004839.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 NM_004839.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 NM_004839.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 NM_004839.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 NM_004839.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 NM_004839.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 NM_004839.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 NM_004839.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 NM_004839.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 NM_004839.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 NM_004839.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 NM_004839.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 NM_004839.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 NM_004839.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 NM_199330.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 NM_199330.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 NM_199330.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 NM_199330.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 NM_199330.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 NM_199330.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 NM_199330.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 NM_199330.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 NM_199330.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 NM_199330.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 NM_199330.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 NM_199330.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 NM_199330.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 NM_199330.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010251.hg.1 NM_199330.2 Refseq
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000304231.12 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000304231.12 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000304231.12 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000304231.12 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000304231.12 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000304231.12 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000304231.12 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000304231.12 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000304231.12 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000304231.12 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000304231.12 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000304231.12 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000304231.12 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000304231.12 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000304231.12 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000450735.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000450735.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000450735.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000450735.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000450735.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000450735.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000450735.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000450735.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000450735.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000450735.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000450735.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000450735.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000450735.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000450735.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000500334.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000500334.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000500334.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000558090.2 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000558090.2 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000558090.2 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000558090.2 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000558552.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000558552.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000558552.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000558552.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000558552.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000558552.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000558817.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000558817.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000558817.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000558817.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000558817.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000558817.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000558817.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000558817.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000560374.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000560374.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000560374.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000560374.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000560374.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000560374.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000561345.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000561345.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000561345.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000561345.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000561345.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000561345.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000561345.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000619240.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000619240.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000619240.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000619367.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000619367.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 ENST00000619367.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010251.hg.1 NM_004839.3 Refseq
Affymetrix Human Clariom D Assay TC1500010251.hg.1 NM_004839.3 Refseq
Affymetrix Human Clariom D Assay TC1500010251.hg.1 NM_004839.3 Refseq
Affymetrix Human Clariom D Assay TC1500010251.hg.1 NM_004839.3 Refseq
Affymetrix Human Clariom D Assay TC1500010251.hg.1 NM_004839.3 Refseq
Affymetrix Human Clariom D Assay TC1500010251.hg.1 NM_004839.3 Refseq
Affymetrix Human Clariom D Assay TC1500010251.hg.1 NM_004839.3 Refseq
Affymetrix Human Clariom D Assay TC1500010251.hg.1 NM_004839.3 Refseq
Affymetrix Human Clariom D Assay TC1500010251.hg.1 NM_004839.3 Refseq
Affymetrix Human Clariom D Assay TC1500010251.hg.1 NM_004839.3 Refseq
Affymetrix Human Clariom D Assay TC1500010251.hg.1 NM_004839.3 Refseq
Affymetrix Human Clariom D Assay TC1500010251.hg.1 NM_004839.3 Refseq
Affymetrix Human Clariom D Assay TC1500010251.hg.1 NM_004839.3 Refseq
Affymetrix Human Clariom D Assay TC1500010251.hg.1 NM_004839.3 Refseq
Affymetrix Human Clariom D Assay TC1500010251.hg.1 NM_199330.2 Refseq
Affymetrix Human Clariom D Assay TC1500010251.hg.1 NM_199330.2 Refseq
Affymetrix Human Clariom D Assay TC1500010251.hg.1 NM_199330.2 Refseq
Affymetrix Human Clariom D Assay TC1500010251.hg.1 NM_199330.2 Refseq
Affymetrix Human Clariom D Assay TC1500010251.hg.1 NM_199330.2 Refseq
Affymetrix Human Clariom D Assay TC1500010251.hg.1 NM_199330.2 Refseq
Affymetrix Human Clariom D Assay TC1500010251.hg.1 NM_199330.2 Refseq
Affymetrix Human Clariom D Assay TC1500010251.hg.1 NM_199330.2 Refseq
Affymetrix Human Clariom D Assay TC1500010251.hg.1 NM_199330.2 Refseq
Affymetrix Human Clariom D Assay TC1500010251.hg.1 NM_199330.2 Refseq
Affymetrix Human Clariom D Assay TC1500010251.hg.1 NM_199330.2 Refseq
Affymetrix Human Clariom D Assay TC1500010251.hg.1 NM_199330.2 Refseq
Affymetrix Human Clariom D Assay TC1500010251.hg.1 NM_199330.2 Refseq
Affymetrix Human Clariom D Assay TC1500010251.hg.1 NM_199330.2 Refseq
Affymetrix Human Clariom D Assay TC1500010251.hg.1 NM_199330.2 Refseq
Human Genome U133 Plus 2.0 Array 1559180_at - -
Human Genome U133 Plus 2.0 Array 1559180_PM_at - -
Affymetrix Human Genome U133 Plus 2.0 Array 1559180_at6 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1559180_at9 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1559180_at11 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1559180_at4 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1559180_at10 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1559180_at3 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1559180_at2 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1559180_at7 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1559180_at1 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1559180_at5 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1559180_at8 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0