This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03655166_s1
See other LOC284513 GE Assays ›
Gene Symbol:
LOC284513
Gene Name:
uncharacterized LOC284513
Gene Aliases:
-
Chromosome Location:
-
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03655166_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC284513
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK096098.1 - 1 3145 73
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB351556.1 - 1 Chr. 1 - 19213488 - 19213717 [-] on Build GRCh38 73
GenBankmRNA DB524658.1 - 1 282 Chr. 1 - 19213258 - 19213714 [+] on Build GRCh38 73
GenBankmRNA AW169311.1 - 1 Chr. 1 - 19213218 - 19213718 [-] on Build GRCh38 73
GenBankmRNA CV575234.1 - 1 279 Chr. 1 - 19213261 - 19213715 [+] on Build GRCh38 73
GenBankmRNA AA758936.1 - 1 Chr. 1 - 19213363 - 19213722 [-] on Build GRCh38 73

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC101927895 LOC101927895 Sense Chr. 1 - 19210386 - 19240704 on NCBI Build GRCh38
Downstream ER membrane protein complex subunit 1 EMC1 CAVIPMR,
KIAA0090
2.0kb Anti-sense Chr. 1 - 19215664 - 19251559 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 38485 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 420830 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 280309 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3856949 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 31634 - -
SurePrint G3 Human GE Microarray G4851A A_33_P3856949 - -
Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P00024174 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15046883 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00024174 - -
Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P00024174 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT HG-U133+ PM Array Plate 1556597_PM_a_at - -
Human Genome U133 Plus 2.0 Array 1556597_PM_a_at - -
Human Genome U133 Plus 2.0 Array 1556597_a_at - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556597_a_at5 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556597_a_at8 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556597_a_at2 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556597_a_at10 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556597_a_at3 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556597_a_at11 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556597_a_at9 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556597_a_at1 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556597_a_at7 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556597_a_at4 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556597_a_at6 - -
Human Genome U95 Set 66428_at - -
Human Genome U95 Set 66428_PM_at - -
Human Genome U95 Set 69863_at - -
Human Genome U95 Set 69863_PM_at - -
Human Genome U95 Set 86180_at - -
Human Genome U95 Set 86180_PM_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0