This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03457523_s1
See other SNORA19 GE Assays ›
Gene Symbol:
SNORA19
Gene Name:
small nucleolar RNA, H/ACA box 19
Gene Aliases:
ACA19
Chromosome Location:
Chr.10: 119060011 - 119060138 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03457523_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SNORA19
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_002917.1 1 54 118
GenBank mRNA AJ609435.1 - 1 54 118

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream nanos C2HC-type zinc finger 1 NANOS1 EC_Rep1a,
NOS-1,
NOS1,
SPGF12,
ZC2HC12A
-25.7kb Anti-sense Chr. 10 - 119029716 - 119033732 on NCBI Build GRCh38
Downstream family with sequence similarity 45 member A FAM45A 43.9kb Anti-sense Chr. 10 - 119104065 - 119137984 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000011989.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10001699.hg.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3924 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3923 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910077 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07519635 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910069 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910096 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07519617 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07519640 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910061 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910067 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910072 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910098 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910082 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16389246 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910070 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910097 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910085 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910079 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910076 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07519618 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910092 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910081 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910065 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07519634 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910068 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07519631 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910094 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910058 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910095 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16389248 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910089 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910087 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07519604 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910056 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07519627 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07519628 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07519602 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07519631 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910068 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910072 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910085 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07519640 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07519627 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910082 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910098 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910065 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910067 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07519635 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910097 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910089 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07519602 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910096 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910061 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910094 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07519604 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07519634 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910095 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16389248 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910056 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910070 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07519628 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07519617 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07519618 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910081 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16389246 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910077 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910079 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910069 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910092 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910058 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910076 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27910087 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07519628 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07519603 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07519641 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07519611 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07519602 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07519609 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07519616 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07519626 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07519632 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07519638 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07519618 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07519647 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07519630 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07519606 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07519600 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07519644 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07519614 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07519637 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07519615 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07519640 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07519608 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07519634 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07519607 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07519621 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07519642 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07519624 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.0 ST Array NR_002917_at - -
Affymetrix Human Gene 1.0 ST Array 7936638 - -
Affymetrix Human Gene 1.1 ST Array 7936637 - -
Affymetrix Human Genome U219 Array 11763919_PM_at - -
Affymetrix Human Genome U219 Array 11763919_at - -
Affymetrix Human Transcriptome Array 2.0 TC1000011989.hg.1 ENST00000384737.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011989.hg.1 ENST00000384737.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011989.hg.1 ENST00000626927.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011989.hg.1 NR_002917.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011989.hg.1 NR_002917.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10022378.hg.1 ENST00000384737.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10022378.hg.1 ENST00000462527.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10022378.hg.1 NR_002917.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10022378.hg.1 TC10001699.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1000182055.hg.1 ENST00000384737.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000182055.hg.1 NR_002917.1 Refseq
Affymetrix Human Clariom D Assay PSR1000182055.hg.1 ENST00000384737.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000182055.hg.1 NR_002917.1 Refseq
Affymetrix Human Clariom D Assay PSR1000182055.hg.1 TC1000011989.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000011989.hg.1 ENST00000384737.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011989.hg.1 ENST00000384737.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011989.hg.1 ENST00000626927.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011989.hg.1 NR_002917.1 Refseq
Affymetrix Human Clariom D Assay TC1000011989.hg.1 NR_002917.1 Refseq
Affymetrix Human Clariom D Assay PSR1000182054.hg.1 ENST00000384737.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000182054.hg.1 ENST00000626927.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000182054.hg.1 NR_002917.1 Refseq
Affymetrix Human Clariom D Assay PSR1000182054.hg.1 ENST00000384737.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000182054.hg.1 ENST00000626927.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000182054.hg.1 NR_002917.1 Refseq
Affymetrix Human Clariom D Assay PSR1000182054.hg.1 TC1000011989.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3242748 NR_002917.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_3242748 AJ609435.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3242748 NR_002917.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3242748 AJ609435.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3242748 NR_002917.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3242748 AJ609435.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3242748 NR_002917.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3242748 AJ609435.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0