This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03309805_s1
See other SCARNA10 GE Assays ›
Gene Symbol:
SCARNA10
Gene Name:
small Cajal body-specific RNA 10
Gene Aliases:
U85
Chromosome Location:
Chr.12: 6510222 - 6510551 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03309805_s1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SCARNA10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_004387.1 1 254 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200006628.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12000083.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ArrayStar Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3363120 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4301 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4302 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 9032 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3363120 - -
SurePrint G3 Human GE Microarray G4851A A_33_P3363120 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08176113 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28605020 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08176111 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28605017 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08176114 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28605023 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28605015 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28605019 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28605021 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28605019 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28605021 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08176111 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28605015 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28605023 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08176114 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28605017 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08176113 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28605020 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08176113 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08176109 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08176105 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08176111 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.0 ST Array NR_004387_at - -
Affymetrix Human Gene 1.0 ST Array 7953384 - -
Affymetrix Human Gene 1.1 ST Array 7953383 - -
Affymetrix Human Genome U219 Array 11757260_PM_at - -
Affymetrix Human Genome U219 Array 11763955_PM_at - -
Affymetrix Human Genome U219 Array 11757260_at - -
Affymetrix Human Genome U219 Array 11763955_at - -
Affymetrix Human Transcriptome Array 2.0 PSR12000901.hg.1 ENST00000459255.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12000901.hg.1 NR_004387.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12000901.hg.1 TC12000083.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR12000902.hg.1 ENST00000459255.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12000902.hg.1 ENST00000541782.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12000902.hg.1 NR_004387.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12000902.hg.1 TC12000083.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR12000903.hg.1 ENST00000459255.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12000903.hg.1 NR_004387.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12000903.hg.1 TC12000083.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1200006628.hg.1 ENST00000459255.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006628.hg.1 ENST00000459255.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006628.hg.1 ENST00000459255.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006628.hg.1 ENST00000541782.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006628.hg.1 NR_004387.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006628.hg.1 NR_004387.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006628.hg.1 NR_004387.1 Refseq
Affymetrix Human Clariom D Assay PSR1200145738.hg.1 ENST00000459255.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200145738.hg.1 ENST00000541782.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200145738.hg.1 NR_004387.1 Refseq
Affymetrix Human Clariom D Assay PSR1200145738.hg.1 ENST00000459255.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200145738.hg.1 ENST00000541782.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200145738.hg.1 NR_004387.1 Refseq
Affymetrix Human Clariom D Assay PSR1200145738.hg.1 TC1200006628.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200145737.hg.1 ENST00000459255.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200145737.hg.1 NR_004387.1 Refseq
Affymetrix Human Clariom D Assay PSR1200145737.hg.1 ENST00000459255.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200145737.hg.1 NR_004387.1 Refseq
Affymetrix Human Clariom D Assay PSR1200145737.hg.1 TC1200006628.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200145739.hg.1 ENST00000459255.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200145739.hg.1 NR_004387.1 Refseq
Affymetrix Human Clariom D Assay PSR1200145739.hg.1 ENST00000459255.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200145739.hg.1 NR_004387.1 Refseq
Affymetrix Human Clariom D Assay PSR1200145739.hg.1 TC1200006628.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1200006628.hg.1 ENST00000459255.1 Ensembl
Affymetrix Human Clariom D Assay TC1200006628.hg.1 ENST00000459255.1 Ensembl
Affymetrix Human Clariom D Assay TC1200006628.hg.1 ENST00000459255.1 Ensembl
Affymetrix Human Clariom D Assay TC1200006628.hg.1 ENST00000541782.1 Ensembl
Affymetrix Human Clariom D Assay TC1200006628.hg.1 NR_004387.1 Refseq
Affymetrix Human Clariom D Assay TC1200006628.hg.1 NR_004387.1 Refseq
Affymetrix Human Clariom D Assay TC1200006628.hg.1 NR_004387.1 Refseq
Array Name Probe IDs Targets Target Type
Arraystar Human LncRNA microarray V2.0 ASHG19A3A048271 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A048271 - -
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3246209 NR_004387.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3246209 NR_004387.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3246209 NR_004387.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3246209 NR_004387.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0