This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03309514_s1
See other RMRP GE Assays ›
Gene Symbol:
RMRP
Gene Name:
RNA component of mitochondrial RNA processing endoribonuclease
Gene Aliases:
CHH, NME1, RMRPR, RRP2
Chromosome Location:
Chr.9: 35657751 - 35658018 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03309514_s1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RMRP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_003051.3 1 56 105
GenBank mRNA DW422801.1 - 1 72 105
DW423815.1 - 1 72 105

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900009947.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09001056.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Inventoried Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0900009947.hg.1 ENST00000363046.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009947.hg.1 ENST00000602361.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009947.hg.1 NR_003051.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0