This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03309321_s1
See other ASAP1-IT1 GE Assays ›
Gene Symbol:
ASAP1-IT1
Gene Name:
ASAP1 intronic transcript 1
Gene Aliases:
ASAP1-IT, ASAP1IT, ASAP1IT1, DDEF1IT1, HSPC054, NCRNA00050
Chromosome Location:
Chr.8: 130295355 - 130296533 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03309321_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ASAP1-IT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_002765.2 1 556 135
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AF150205.1 - 1 554 Chr. 8 - 130295355 - 130296523 [-] on Build GRCh38 134

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ASAP1 intronic transcript 2 ASAP1-IT2 -210.6kb Sense Chr. 8 - 130082738 - 130084768 on NCBI Build GRCh38
Downstream adenylate cyclase 8 ADCY8 AC8,
ADCY3,
HBAC1
483.8kb Sense Chr. 8 - 130780300 - 131041604 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0