Assay Details
Gene Symbol: PIN4P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_003571.1 | 1 | 1846 | 125 | ||
GenBank mRNA | AK055299.1 | - | 1 | 1846 | 125 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | FERM domain containing 5 | FRMD5 | Sense | Chr. 15 - 43870761 - 44195294 on NCBI Build GRCh38 | ||
Downstream | cancer susceptibility candidate 4 | CASC4 | H63 | 410.4kb | Anti-sense | Chr. 15 - 44288616 - 44415761 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC15001290.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4200 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4201 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3235987 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30112489 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621027 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621029 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16785288 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30112486 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621054 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621073 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621039 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621020 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621008 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30112501 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621022 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621031 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621014 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16785293 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30112540 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621075 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621056 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621050 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621012 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621049 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16785291 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621030 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621068 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30112550 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621061 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621044 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621057 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621035 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30112494 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30112505 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30112534 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621002 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30112536 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621018 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621053 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621034 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621003 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30112525 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621078 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621064 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621036 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30112521 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621076 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621023 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621006 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621063 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621009 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16785289 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621070 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30112519 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621043 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621037 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621017 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16785291 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621036 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30112550 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621057 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621037 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621020 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621076 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621064 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621027 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30112519 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30112540 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621030 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30112525 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621075 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621050 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621043 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621035 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621070 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30112536 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621029 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30112494 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16785293 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621022 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621049 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30112489 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621053 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621012 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621054 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621061 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621031 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621034 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621014 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621003 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621056 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621009 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621039 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621006 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621017 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621023 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30112505 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621078 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30112534 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16785288 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30112486 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621063 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621073 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16785289 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621044 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621008 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30112521 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621018 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621002 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30112501 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09621068 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621059 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621040 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621068 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621046 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621076 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621048 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621034 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16785288 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621042 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621036 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621011 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621043 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621003 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621081 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621064 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621084 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621020 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621014 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621033 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621016 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621006 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621022 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621039 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16785289 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621078 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621027 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621062 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621030 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621070 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621044 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621080 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16785291 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621056 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621053 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16785294 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621073 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621050 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621031 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621002 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621018 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09621009 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1500009239.hg.1 | ENST00000451079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009239.hg.1 | NR_003571.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009239.hg.1 | NR_003571.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011507.hg.1 | ENST00000416056.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011708.hg.1 | ENST00000446145.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011708.hg.1 | ENST00000446145.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011708.hg.1 | ENST00000446145.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15012335.hg.1 | NR_003571.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15012335.hg.1 | TC15001290.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1500009239.hg.1 | ENST00000451079.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009239.hg.1 | NR_003571.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009239.hg.1 | NR_003571.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500167697.hg.1 | NR_003571.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500167697.hg.1 | NR_003571.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500167697.hg.1 | TC1500009239.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100011507.hg.1 | ENST00000416056.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011708.hg.1 | ENST00000446145.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011708.hg.1 | ENST00000446145.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011708.hg.1 | ENST00000446145.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500167696.hg.1 | ENST00000451079.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500167696.hg.1 | NR_003571.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500167696.hg.1 | ENST00000451079.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500167696.hg.1 | NR_003571.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500167696.hg.1 | TC1500009239.hg.1 | Clariom D TC |
AffymetrixPathwork Diagnostics Pathchip | pw500084 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A052092 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A052092 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 347936 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_2134855 | BC070288.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2134855 | BC111394.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2134855 | BU952002.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2134855 | NM_006223.3 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2134855 | NR_033187.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2134855 | BC070288.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2134855 | BC111394.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2134855 | BU952002.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2134855 | NM_006223.3 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2134855 | NR_033187.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2134855 | BC070288.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2134855 | BC111394.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2134855 | BU952002.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2134855 | NM_006223.3 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2134855 | NR_033187.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2134855 | BC070288.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2134855 | BC111394.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2134855 | BU952002.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2134855 | NM_006223.3 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2134855 | NR_033187.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2134855 | BC070288.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2134855 | BC111394.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2134855 | BU952002.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2134855 | NM_006223.3 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2134855 | NR_033187.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2134855 | BC070288.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2134855 | BC111394.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2134855 | BU952002.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2134855 | NM_006223.3 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2134855 | NR_033187.1 | human_transcriptome |
