This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02567210_s1
See other BAZ1A GE Assays ›
Gene Symbol:
BAZ1A
Gene Name:
bromodomain adjacent to zinc finger domain 1A
Gene Aliases:
ACF1, WALp1, WCRF180, hACF1
Chromosome Location:
Chr.14: 34752731 - 34875647 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs02567210_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BAZ1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013448.2 NP_038476.2 13 2135 63
XM_011536374.1 XP_011534676.1 12 1434 63
XM_011536376.2 XP_011534678.2 3 262 63
GenBank mRNA AF213467.1 - 13 1733 63
AF221130.1 - 12 1567 63

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene BAZ1A

Gene Ontology Categories:

Function(s) Process(es)

DNA-dependent DNA replication
chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
histone acetyltransferase activity
protein binding
zinc ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0