This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02559514_s1
See other SMARCC2 GE Assays ›
Gene Symbol:
SMARCC2
Gene Name:
SWI/SNF related, matrix associated, actin dependent regulator of chromatin subfamily c member 2
Gene Aliases:
BAF170, CRACC2, Rsc8
Chromosome Location:
Chr.12: 56161852 - 56189567 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs02559514_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SMARCC2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001130420.1 NP_001123892.1 11 1143 90
NM_003075.3 NP_003066.2 11 1143 90
NM_139067.2 NP_620706.1 11 1143 90
XM_005269101.1 XP_005269158.1 11 1080 90
XM_005269102.1 XP_005269159.1 11 1080 90
XM_005269103.1 XP_005269160.1 11 1080 90
XM_005269104.1 XP_005269161.1 11 1080 90
XM_011538693.2 XP_011536995.1 10 989 90
XM_017019884.1 XP_016875373.1 11 1080 90
XM_017019885.1 XP_016875374.1 11 1080 90
XM_017019886.1 XP_016875375.1 11 1080 90
XM_017019887.1 XP_016875376.1 10 967 90
GenBank mRNA AB209006.1 - 11 1143 90
AB209293.1 - 11 1052 90
AK293896.1 - 11 1030 90
AK315153.1 - 11 1043 90
BC013045.1 - 11 1069 4180388 90
BC026222.2 - 11 1062 4395012 90
BT009924.1 - 11 1037 90
U66616.1 - 11 1059 90

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SMARCC2

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
nucleosome disassembly
chromatin remodeling
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
covalent chromatin modification
regulation of transcription from RNA polymerase II promoter involved in forebrain neuron fate commitment
ATP-dependent chromatin remodeling
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
transcription coactivator activity
protein binding
nucleosomal DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0