This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01895061_u1
See other POU5F1P3 GE Assays ›
Gene Symbol:
POU5F1P3 POU5F1 POU5F1P4
Gene Name:
POU class 5 homeobox 1 pseudogene 3
POU class 5 homeobox 1
POU class 5 homeobox 1 pseudogene 4
Gene Aliases:
OCT4-pg3, OTF3L, POU5F1L, POU5FLC12
OCT3, OCT4, OTF-3, OTF3, OTF4, Oct-3, Oct-4
Oct4-pg4, POU5FLC1
Chromosome Location:
Chr.12: 8133769 - 8134852 on Build GRCh38
Chromosome Location:
Chr.6: 31164337 - 31170693 on Build GRCh38
Chromosome Location:
Chr.1: 155433180 - 155434262 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
-
-
Assay ID Hs01895061_u1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: POU5F1P3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_036440.1 1 130
GenBank mRNA AF268617.1 - 1 395 130
FM177983.1 - 1 274 130
FM177984.1 - 1 274 130
FM177985.1 - 1 274 130
FM177987.1 - 1 274 130
FM177988.1 - 1 274 130
FM177989.1 - 1 274 130
FM177990.1 - 1 274 130
FM177991.1 - 1 274 130
FM177992.1 - 1 274 130
FM177995.1 - 1 274 130
FM177997.1 - 1 274 130
FM177999.1 - 1 274 130
GU480832.1 - 1 396 130
GU480837.1 - 1 396 130
GU480838.1 - 1 396 130
GU480846.1 - 1 396 130
GU480876.1 - 1 396 130
GU480877.1 - 1 396 130
GU480887.1 - 1 396 130
GU480888.1 - 1 397 130
GU480891.1 - 1 397 130
GU480892.1 - 1 396 130
GU480897.1 - 1 396 130
GU480901.1 - 1 395 130
GU480903.1 - 1 396 130
GU480907.1 - 1 396 130
GU480909.1 - 1 396 130
GU480915.1 - 1 130
HQ122694.1 - 1 130
HQ122722.1 - 1 396 130
HQ122724.1 - 1 396 130
HQ122725.1 - 1 396 130
HQ122726.1 - 1 396 130
HQ122728.1 - 1 396 130
HQ122729.1 - 1 396 130
HQ122730.1 - 1 396 130
HQ122753.1 - 1 396 130
HQ122754.1 - 1 396 130
HQ122791.1 - 1 396 130
HQ122792.1 - 1 396 130
HQ122793.1 - 1 396 130
HQ122794.1 - 1 396 130
HQ122795.1 - 1 396 130
HQ122796.1 - 1 396 130
HQ122828.1 - 1 396 130
HQ122829.1 - 1 396 130
HQ122830.1 - 1 396 130
HQ122831.1 - 1 396 130
Gene Symbol: POU5F1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002701.5 NP_002692.2 1 130
GenBank mRNA BC117435.1 - 1 40125986 130
BC117437.1 - 1 40125988 130
BQ278003.1 - 130
CD247724.1 - 1 130
FM177951.1 - 2 130
FM177952.1 - 1 130
FM177953.1 - 1 130
FM177954.1 - 1 130
FM177955.1 - 1 130
FM177956.1 - 1 130
FM177957.1 - 2 130
FM177958.1 - 2 130
FM177960.1 - 1 130
FM177961.1 - 1 130
FM177962.1 - 1 130
FM177963.1 - 1 130
FM177964.1 - 2 130
FM177965.1 - 1 130
FM177968.1 - 1 130
GU480808.1 - 1 130
GU480809.1 - 1 130
GU480811.1 - 1 130
GU480812.1 - 1 130
GU480813.1 - 1 130
GU480815.1 - 1 130
GU480816.1 - 1 130
GU480817.1 - 1 130
GU480818.1 - 1 130
GU480819.1 - 1 130
GU480820.1 - 1 130
GU480821.1 - 1 130
GU480822.1 - 1 130
GU480824.1 - 1 130
GU480825.1 - 1 130
GU480826.1 - 1 130
GU480828.1 - 1 130
GU480829.1 - 1 130
GU480830.1 - 1 130
HQ122658.1 - 1 130
HQ122660.1 - 1 130
HQ122661.1 - 1 130
HQ122662.1 - 1 130
HQ122663.1 - 1 130
HQ122664.1 - 1 130
HQ122665.1 - 1 130
HQ122666.1 - 1 130
HQ122667.1 - 1 130
HQ122668.1 - 1 130
HQ122669.1 - 1 130
HQ122670.1 - 1 130
HQ122671.1 - 1 130
HQ122672.1 - 1 130
HQ122673.1 - 1 130
HQ122674.1 - 1 130
HQ122675.1 - 1 130
Z11898.1 - 1 130
Gene Symbol: POU5F1P4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA HQ122756.1 - 1 130
HQ122860.1 - 1 130

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600011406.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06001521.hg.1, TC6_cox_hap2000153.hg.1, TC6_dbb_hap3000142.hg.1, TC6_mann_hap4000130.hg.1, TC6_mcf_hap5000130.hg.1, TC6_qbl_hap6000143.hg.1, TC6_ssto_hap7000125.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700 Inventoried Amplicon spans exons and probe does not span exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene POU5F1P3
Click here to find siRNAs for Gene POU5F1
Click here to find siRNAs for Gene POU5F1P4

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
endodermal cell fate specification
blastocyst development
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
response to wounding
anatomical structure morphogenesis
regulation of asymmetric cell division
regulation of gene expression
somatic stem cell population maintenance
mRNA transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
negative regulation of gene silencing by miRNA
transcription regulatory region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
ubiquitin protein ligase binding
miRNA binding
sequence-specific DNA binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR6_mann_hap4002175.hg.1 NM_001173531.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_mann_hap4002175.hg.1 NM_002701.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_mann_hap4002175.hg.1 NM_203289.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_mann_hap4002175.hg.1 TC6_mann_hap4000130.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_dbb_hap3002592.hg.1 NM_001173531.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_dbb_hap3002592.hg.1 NM_002701.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_dbb_hap3002592.hg.1 NM_203289.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_dbb_hap3002592.hg.1 TC6_dbb_hap3000142.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR06017287.hg.1 ENST00000259915.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06017287.hg.1 ENST00000441888.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06017287.hg.1 ENST00000448657.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06017287.hg.1 ENST00000471529.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06017287.hg.1 ENST00000512818.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06017287.hg.1 NM_001173531.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06017287.hg.1 NM_002701.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06017287.hg.1 NM_203289.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06017287.hg.1 TC06001521.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2002656.hg.1 NM_001173531.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2002656.hg.1 NM_002701.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2002656.hg.1 NM_203289.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2002656.hg.1 TC6_cox_hap2000153.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6002632.hg.1 NM_001173531.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6002632.hg.1 NM_002701.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6002632.hg.1 NM_203289.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6002632.hg.1 TC6_qbl_hap6000143.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR06017291.hg.1 ENST00000259915.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06017291.hg.1 ENST00000448657.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06017291.hg.1 NM_002701.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06017291.hg.1 TC06001521.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_ssto_hap7002316.hg.1 NM_001173531.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_ssto_hap7002316.hg.1 NM_002701.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_ssto_hap7002316.hg.1 NM_203289.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_ssto_hap7002316.hg.1 TC6_ssto_hap7000125.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_mcf_hap5002323.hg.1 NM_001173531.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_mcf_hap5002323.hg.1 NM_002701.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_mcf_hap5002323.hg.1 NM_203289.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_mcf_hap5002323.hg.1 TC6_mcf_hap5000130.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0600177023.hg.1 ENST00000259915.12 Ensembl
Affymetrix Human Clariom D Assay PSR0600177023.hg.1 ENST00000461401.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600177023.hg.1 ENST00000619340.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600177023.hg.1 ENST00000259915.12 Ensembl
Affymetrix Human Clariom D Assay PSR0600177023.hg.1 ENST00000461401.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600177023.hg.1 ENST00000619340.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600177023.hg.1 TC0600011406.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0600177026.hg.1 ENST00000259915.12 Ensembl
Affymetrix Human Clariom D Assay PSR0600177026.hg.1 ENST00000461401.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600177026.hg.1 ENST00000259915.12 Ensembl
Affymetrix Human Clariom D Assay PSR0600177026.hg.1 ENST00000461401.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600177026.hg.1 TC0600011406.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0600177024.hg.1 ENST00000259915.12 Ensembl
Affymetrix Human Clariom D Assay PSR0600177024.hg.1 ENST00000461401.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600177024.hg.1 ENST00000619340.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600177024.hg.1 ENST00000259915.12 Ensembl
Affymetrix Human Clariom D Assay PSR0600177024.hg.1 ENST00000461401.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600177024.hg.1 ENST00000619340.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600177024.hg.1 TC0600011406.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 193216 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0