Assay Details
Gene Symbol: APOL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030882.3 | NP_112092.2 | 5 | 1100 | 128 | |
NM_145637.2 | NP_663612.2 | 6 | 1241 | 128 | ||
XM_011530074.1 | XP_011528376.1 | 6 | 1807 | 128 | ||
XM_011530075.1 | XP_011528377.1 | 7 | 1813 | 128 | ||
XM_011530076.2 | XP_011528378.1 | 6 | 1118 | 128 | ||
XM_011530077.2 | XP_011528379.1 | 5 | 1196 | 128 | ||
XM_011530078.1 | XP_011528380.1 | 7 | 1907 | 128 | ||
XM_017028724.1 | XP_016884213.1 | 6 | 1115 | 128 | ||
GenBank mRNA | AB208990.1 | - | 5 | 1000 | 128 | |
AF305225.1 | - | 5 | 984 | 128 | ||
AF305429.1 | - | 5 | 963 | 128 | ||
AF324223.1 | - | 5 | 1100 | 128 | ||
AF324224.1 | - | 6 | 1241 | 128 | ||
AK056938.1 | - | 5 | 1032 | 128 | ||
AK303978.1 | - | 6 | 1109 | 128 | ||
BC004395.1 | - | 5 | 968 | 3636011 | 128 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000249066.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000249066.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000249066.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000249066.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000249066.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000249066.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000249066.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000249066.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000249066.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000249066.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000249066.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000249066.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000358502.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000358502.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000358502.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000358502.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000358502.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000358502.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000358502.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000358502.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000358502.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000358502.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000358502.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000358502.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000451256.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000451256.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000451256.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000451256.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000451256.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000451256.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000451256.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000451256.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000451256.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000454728.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000454728.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000454728.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000454728.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000454728.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000454728.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000454728.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000473549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000473549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000473549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000473549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000473549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000473549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000473549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000476579.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000476579.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000476579.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000476579.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000476579.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000476579.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000476579.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000484830.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000484830.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000484830.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000484830.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000484830.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000489186.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000489186.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000489186.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000489186.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000489186.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000489186.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000529194.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000529194.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000529194.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000529194.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000529194.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000529194.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | ENST00000529194.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | NM_030882.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | NM_030882.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | NM_030882.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | NM_030882.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | NM_030882.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | NM_030882.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | NM_030882.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | NM_030882.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | NM_030882.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | NM_030882.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | NM_030882.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | NM_030882.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | NM_145637.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | NM_145637.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | NM_145637.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | NM_145637.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | NM_145637.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | NM_145637.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | NM_145637.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | NM_145637.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | NM_145637.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | NM_145637.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | NM_145637.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008592.hg.1 | NM_145637.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000249066.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000249066.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000249066.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000249066.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000249066.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000249066.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000249066.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000249066.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000249066.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000249066.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000249066.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000249066.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000358502.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000358502.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000358502.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000358502.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000358502.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000358502.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000358502.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000358502.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000358502.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000358502.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000358502.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000358502.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000451256.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000451256.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000451256.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000451256.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000451256.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000451256.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000451256.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000451256.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000451256.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000454728.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000454728.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000454728.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000454728.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000454728.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000454728.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000454728.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000473549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000473549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000473549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000473549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000473549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000473549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000473549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000476579.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000476579.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000476579.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000476579.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000476579.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000476579.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000476579.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000484830.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000484830.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000484830.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000484830.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000484830.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000489186.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000489186.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000489186.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000489186.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000489186.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000489186.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000529194.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000529194.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000529194.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000529194.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000529194.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000529194.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | ENST00000529194.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | NM_030882.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | NM_030882.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | NM_030882.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | NM_030882.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | NM_030882.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | NM_030882.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | NM_030882.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | NM_030882.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | NM_030882.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | NM_030882.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | NM_030882.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | NM_030882.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | NM_145637.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | NM_145637.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | NM_145637.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | NM_145637.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | NM_145637.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | NM_145637.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | NM_145637.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | NM_145637.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | NM_145637.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | NM_145637.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | NM_145637.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008592.hg.1 | NM_145637.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200161862.hg.1 | ENST00000249066.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200161862.hg.1 | ENST00000358502.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200161862.hg.1 | ENST00000451256.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200161862.hg.1 | NM_030882.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200161862.hg.1 | NM_145637.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200161862.hg.1 | ENST00000249066.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200161862.hg.1 | ENST00000358502.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200161862.hg.1 | ENST00000451256.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200161862.hg.1 | NM_030882.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200161862.hg.1 | NM_145637.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200161862.hg.1 | TC2200008592.hg.1 | Clariom D TC |
