Assay Details
Gene Symbol: POU5F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002701.5 | NP_002692.2 | 1 | 60 | 162 | |
GenBank mRNA | BQ278003.1 | - | 43 | 162 | ||
CD247724.1 | - | 1 | 83 | 162 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_002701.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
endodermal cell fate specification
blastocyst development
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
response to wounding
anatomical structure morphogenesis
regulation of asymmetric cell division
regulation of gene expression
somatic stem cell population maintenance
mRNA transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
negative regulation of gene silencing by miRNA
transcription regulatory region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
ubiquitin protein ligase binding
miRNA binding
sequence-specific DNA binding
poly(A) RNA binding
negative regulation of transcription from RNA polymerase II promoter
endodermal cell fate specification
blastocyst development
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
response to wounding
anatomical structure morphogenesis
regulation of asymmetric cell division
regulation of gene expression
somatic stem cell population maintenance
mRNA transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
negative regulation of gene silencing by miRNA
transcription regulatory region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
ubiquitin protein ligase binding
miRNA binding
sequence-specific DNA binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR06017293.hg.1 | ENST00000259915.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06017293.hg.1 | ENST00000448657.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06017293.hg.1 | NM_002701.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06017293.hg.1 | TC06001521.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002327.hg.1 | NM_002701.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002327.hg.1 | TC6_mcf_hap5000130.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002636.hg.1 | NM_002701.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002636.hg.1 | TC6_qbl_hap6000143.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06017294.hg.1 | ENST00000259915.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06017294.hg.1 | NM_002701.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06017294.hg.1 | TC06001521.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4002179.hg.1 | NM_002701.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4002179.hg.1 | TC6_mann_hap4000130.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000259915.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000259915.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000259915.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000259915.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000259915.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000259915.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000259915.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000259915.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000259915.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000259915.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000441888.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000441888.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000441888.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000441888.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000441888.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000441888.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000441888.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000441888.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000461401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000461401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000461401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000461401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000461401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000471529.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000471529.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000471529.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000471529.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000471529.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000471529.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000471529.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000471529.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000471529.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000512818.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000512818.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000512818.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000512818.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000512818.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000512818.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000512818.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000512818.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000513407.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000513407.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000513407.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000513407.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000513407.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000513407.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000513407.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000513407.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000513407.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000513407.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000513407.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000606567.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000606567.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000606567.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000606567.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000606567.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000606567.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000606567.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000606567.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000619340.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000619340.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000619340.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000619340.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000619340.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000619340.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000619340.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000620031.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000620031.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000620031.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000620031.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000620031.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000620031.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000620031.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000620031.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011406.hg.1 | ENST00000620031.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002596.hg.1 | NM_002701.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002596.hg.1 | TC6_dbb_hap3000142.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002660.hg.1 | NM_002701.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002660.hg.1 | TC6_cox_hap2000153.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002320.hg.1 | NM_002701.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002320.hg.1 | TC6_ssto_hap7000125.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000259915.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000259915.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000259915.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000259915.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000259915.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000259915.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000259915.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000259915.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000259915.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000259915.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000441888.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000441888.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000441888.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000441888.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000441888.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000441888.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000441888.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000441888.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000461401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000461401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000461401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000461401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000461401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000471529.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000471529.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000471529.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000471529.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000471529.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000471529.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000471529.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000471529.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000471529.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000512818.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000512818.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000512818.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000512818.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000512818.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000512818.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000512818.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000512818.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000513407.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000513407.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000513407.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000513407.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000513407.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000513407.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000513407.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000513407.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000513407.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000513407.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000513407.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000606567.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000606567.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000606567.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000606567.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000606567.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000606567.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000606567.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000606567.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000619340.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000619340.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000619340.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000619340.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000619340.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000619340.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000619340.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000620031.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000620031.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000620031.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000620031.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000620031.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000620031.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000620031.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000620031.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011406.hg.1 | ENST00000620031.4 | Ensembl |
