Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014659.hg.1 |
ENST00000421331.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014659.hg.1 |
ENST00000421331.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014659.hg.1 |
ENST00000421331.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014659.hg.1 |
ENST00000421331.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014659.hg.1 |
ENST00000421331.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014659.hg.1 |
ENST00000597757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014659.hg.1 |
ENST00000597757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014659.hg.1 |
ENST00000597757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014659.hg.1 |
ENST00000597757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014659.hg.1 |
NR_103535.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014659.hg.1 |
NR_103535.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014659.hg.1 |
NR_103535.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014659.hg.1 |
NR_103535.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014659.hg.1 |
NR_103535.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014659.hg.1 |
ENST00000421331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014659.hg.1 |
ENST00000421331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014659.hg.1 |
ENST00000421331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014659.hg.1 |
ENST00000421331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014659.hg.1 |
ENST00000421331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014659.hg.1 |
ENST00000597757.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014659.hg.1 |
ENST00000597757.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014659.hg.1 |
ENST00000597757.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014659.hg.1 |
ENST00000597757.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014659.hg.1 |
NR_103535.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014659.hg.1 |
NR_103535.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014659.hg.1 |
NR_103535.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014659.hg.1 |
NR_103535.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014659.hg.1 |
NR_103535.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100209357.hg.1 |
ENST00000421331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100209357.hg.1 |
NR_103535.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100209357.hg.1 |
ENST00000421331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100209357.hg.1 |
NR_103535.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100209357.hg.1 |
TC0100014659.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100209358.hg.1 |
ENST00000421331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100209358.hg.1 |
NR_103535.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100209358.hg.1 |
ENST00000421331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100209358.hg.1 |
NR_103535.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100209358.hg.1 |
TC0100014659.hg.1 |
Clariom D TC |