This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01651051_m1
See other CFAP54 GE Assays ›
Gene Symbol:
CFAP54
Gene Name:
cilia and flagella associated protein 54
Gene Aliases:
C12orf55, C12orf63
Chromosome Location:
Chr.12: 96489566 - 96875555 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01651051_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CFAP54
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001306084.1 NP_001293013.1 45 - 46 6386 177
XM_011539069.2 XP_011537371.1 46 - 47 6586 177
XM_011539070.2 XP_011537372.1 46 - 47 6586 177
XM_011539071.2 XP_011537373.1 46 - 47 6586 177
XM_011539072.2 XP_011537374.1 46 - 47 6586 177
XM_011539073.2 XP_011537375.1 46 - 47 6586 177
XM_011539074.2 XP_011537376.1 46 - 47 6586 177
XM_011539076.2 XP_011537378.1 46 - 47 6586 177
XM_011539077.2 XP_011537379.1 45 - 46 6736 177
XM_011539078.2 XP_011537380.1 46 - 47 6586 177
XM_011539079.1 XP_011537381.1 42 - 43 5832 177
XM_011539081.2 XP_011537383.1 46 - 47 6586 177
XM_011539083.2 XP_011537385.1 46 - 47 6586 177
XM_011539084.2 XP_011537386.1 46 - 47 6586 177
XM_011539085.1 XP_011537387.1 31 - 32 4407 177
XM_017018866.1 XP_016874355.1 46 - 47 6586 177
XM_017018867.1 XP_016874356.1 35 - 36 4838 177
GenBank mRNA AK126100.1 - 12 - 13 1700 177

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200008541.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

structural protein

Gene Ontology Categories:

Function(s)

spermatogenesis
cell differentiation
cilium assembly
cilium movement involved in cell motility

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR1200162652.hg.1 ENST00000342887.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200162652.hg.1 ENST00000524981.8 Ensembl
Affymetrix Human Clariom D Assay PSR1200162652.hg.1 NM_001306084.1 Refseq
Affymetrix Human Clariom D Assay PSR1200162652.hg.1 ENST00000342887.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200162652.hg.1 ENST00000524981.8 Ensembl
Affymetrix Human Clariom D Assay PSR1200162652.hg.1 NM_001306084.1 Refseq
Affymetrix Human Clariom D Assay PSR1200162652.hg.1 TC1200008541.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0