Assay Details
Gene Symbol: RPL23P8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_026673.1 | 1 | 267 | 145 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Sp8 transcription factor | SP8 | BTD | -40.4kb | Anti-sense | Chr. 7 - 20782274 - 20786889 on NCBI Build GRCh38 |
Downstream | long intergenic non-protein coding RNA 1162 | LINC01162 | 7.6kb | Sense | Chr. 7 - 20835431 - 21023148 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07000128.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3295066 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 42437 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3295066 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3295066 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565323 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348643 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565325 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348683 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565320 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348641 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565291 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348672 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565302 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348686 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565296 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348682 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565313 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15915855 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348642 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565300 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348648 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348685 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348664 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565310 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565308 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565309 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348659 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15915853 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565321 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348670 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565307 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348669 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348646 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565319 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348652 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565294 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565303 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348655 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565297 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348675 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348665 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348659 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565323 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348686 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348670 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565291 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565302 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348669 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348665 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348641 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348683 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565310 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15915853 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348646 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348648 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348672 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348655 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565321 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565307 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565297 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565309 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565313 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348643 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565296 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565319 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565300 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15915855 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348642 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348682 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565308 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348675 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565325 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565320 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348664 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565303 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348652 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25565294 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05348685 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348643 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15915853 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348711 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348679 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348686 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348677 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15915859 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15915861 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348698 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348710 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348673 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348644 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348664 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348712 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348663 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15915855 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348701 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348681 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348653 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348655 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348695 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348641 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348665 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348694 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348667 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348658 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348647 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348700 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348715 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348703 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348660 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348709 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348652 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348718 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348675 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348688 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348670 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348649 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348706 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348690 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348661 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348704 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05348684 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8131706 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8131705 | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0700006856.hg.1 | ENST00000405810.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006856.hg.1 | ENST00000405810.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006856.hg.1 | ENST00000405810.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006856.hg.1 | NR_026673.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006856.hg.1 | NR_026673.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006856.hg.1 | NR_026673.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006856.hg.1 | NR_026673.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006856.hg.1 | NR_026673.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07001980.hg.1 | NR_026673.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07001980.hg.1 | TC07000128.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0700147382.hg.1 | NR_026673.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700147382.hg.1 | NR_026673.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700147382.hg.1 | TC0700006856.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700006856.hg.1 | ENST00000405810.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006856.hg.1 | ENST00000405810.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006856.hg.1 | ENST00000405810.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006856.hg.1 | NR_026673.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006856.hg.1 | NR_026673.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006856.hg.1 | NR_026673.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006856.hg.1 | NR_026673.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006856.hg.1 | NR_026673.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700147379.hg.1 | ENST00000405810.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700147379.hg.1 | NR_026673.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700147379.hg.1 | ENST00000405810.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700147379.hg.1 | NR_026673.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700147379.hg.1 | TC0700006856.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700147380.hg.1 | ENST00000405810.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700147380.hg.1 | NR_026673.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700147380.hg.1 | ENST00000405810.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700147380.hg.1 | NR_026673.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700147380.hg.1 | TC0700006856.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700147381.hg.1 | ENST00000405810.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700147381.hg.1 | NR_026673.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700147381.hg.1 | ENST00000405810.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700147381.hg.1 | NR_026673.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700147381.hg.1 | TC0700006856.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700147378.hg.1 | NR_026673.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700147378.hg.1 | NR_026673.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700147378.hg.1 | TC0700006856.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A034034 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A034034 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 206078 | - | - |
