This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01587540_s1
See other HIST2H2AB GE Assays ›
Gene Symbol:
HIST2H2AB
Gene Name:
histone cluster 2, H2ab
Gene Aliases:
H2AB
Chromosome Location:
Chr.1: 149887469 - 149887916 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01587540_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HIST2H2AB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_175065.2 NP_778235.1 1 351 118

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HIST2H2AB

Panther Classification:

Molecular Function -

histone

Gene Ontology Categories:

Function(s) Process(es)

chromatin silencing
biological_process
molecular_function
DNA binding
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0