This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01568993_g1
See other FER1L6-AS1 GE Assays ›
Gene Symbol:
FER1L6-AS1
Gene Name:
FER1L6 antisense RNA 1
Gene Aliases:
C8orf54
Chromosome Location:
Chr.8: 123984138 - 124040782 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01568993_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FER1L6-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_040044.1 3 - 4 522 65
GenBank mRNA AK093040.1 - 3 - 4 522 65
BC137357.1 - 3 - 4 489 9021355 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0800011697.hg.1
This assay targets the same location as HTA transcript cluster(s) TC08001595.hg.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800011697.hg.1 ENST00000518567.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011697.hg.1 ENST00000518567.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011697.hg.1 ENST00000518567.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011697.hg.1 ENST00000518567.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011697.hg.1 ENST00000518567.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011697.hg.1 ENST00000518567.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011697.hg.1 ENST00000518567.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011697.hg.1 ENST00000518567.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011697.hg.1 NR_040044.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011697.hg.1 NR_040044.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011697.hg.1 NR_040044.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011697.hg.1 NR_040044.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011697.hg.1 NR_040044.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011697.hg.1 NR_040044.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011697.hg.1 NR_040044.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011697.hg.1 NR_040044.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR08020871.hg.1 ENST00000518567.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08020871.hg.1 NR_040044.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR08020871.hg.1 TC08001595.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0800011697.hg.1 ENST00000518567.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011697.hg.1 ENST00000518567.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011697.hg.1 ENST00000518567.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011697.hg.1 ENST00000518567.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011697.hg.1 ENST00000518567.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011697.hg.1 ENST00000518567.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011697.hg.1 ENST00000518567.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011697.hg.1 ENST00000518567.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011697.hg.1 NR_040044.1 Refseq
Affymetrix Human Clariom D Assay TC0800011697.hg.1 NR_040044.1 Refseq
Affymetrix Human Clariom D Assay TC0800011697.hg.1 NR_040044.1 Refseq
Affymetrix Human Clariom D Assay TC0800011697.hg.1 NR_040044.1 Refseq
Affymetrix Human Clariom D Assay TC0800011697.hg.1 NR_040044.1 Refseq
Affymetrix Human Clariom D Assay TC0800011697.hg.1 NR_040044.1 Refseq
Affymetrix Human Clariom D Assay TC0800011697.hg.1 NR_040044.1 Refseq
Affymetrix Human Clariom D Assay TC0800011697.hg.1 NR_040044.1 Refseq
Affymetrix Human Clariom D Assay PSR0800179425.hg.1 ENST00000518567.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800179425.hg.1 NR_040044.1 Refseq
Affymetrix Human Clariom D Assay PSR0800179425.hg.1 ENST00000518567.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800179425.hg.1 NR_040044.1 Refseq
Affymetrix Human Clariom D Assay PSR0800179425.hg.1 TC0800011697.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0