This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01562427_m1
See other PMS2 GE Assays ›
Gene Symbol:
PMS2
Gene Name:
PMS1 homolog 2, mismatch repair system component
Gene Aliases:
HNPCC4, MLH4, PMS2CL, PMSL2
Chromosome Location:
Chr.7: 5970925 - 6009106 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01562427_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PMS2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000535.6 NP_000526.2 7 - 8 891 107
NM_001322003.1 NP_001308932.1 7 - 8 886 107
NM_001322004.1 NP_001308933.1 6 - 7 751 107
NM_001322005.1 NP_001308934.1 7 - 8 1003 107
NM_001322006.1 NP_001308935.1 7 - 8 891 107
NM_001322007.1 NP_001308936.1 6 - 7 788 107
NM_001322008.1 NP_001308937.1 5 - 6 648 107
NM_001322009.1 NP_001308938.1 7 - 8 998 107
NM_001322010.1 NP_001308939.1 6 - 7 751 107
NM_001322011.1 NP_001308940.1 7 - 8 842 107
NM_001322012.1 NP_001308941.1 7 - 8 837 107
NM_001322013.1 NP_001308942.1 6 - 7 718 107
NM_001322014.1 NP_001308943.1 7 - 8 891 107
NM_001322015.1 NP_001308944.1 7 - 8 1061 107
NR_136154.1 7 - 8 891 107
GenBank mRNA AB103082.1 - 7 - 8 804 107
AB103086.1 - 4 - 5 486 107
AK294661.1 - 6 - 7 741 107
AK312390.1 - 7 - 8 840 107
BC008400.1 - 7 - 8 849 107
BC031832.1 - 7 - 8 855 107
BC093921.1 - 7 - 8 826 7939766 107
BP251389.1 - 3 - 4 361 107
U14658.1 - 7 - 8 806 107

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700010182.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07001124.hg.1

Additional Information:

This assay is in the 5'UTR region of the NM_001322011.1,NM_001322012.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PMS2

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

mismatch repair
somatic hypermutation of immunoglobulin genes
response to drug
nucleic acid phosphodiester bond hydrolysis
DNA binding
single-stranded DNA binding
endonuclease activity
protein binding
ATP binding
ATPase activity
single base insertion or deletion binding
MutSalpha complex binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000380416.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000380416.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000380416.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000380416.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000380416.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000380416.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000380416.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000415839.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000415839.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000415839.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000415839.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000415839.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000415839.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000415839.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000469652.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000469652.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0