This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01551773_g1
See other LGR4 GE Assays ›
Gene Symbol:
LGR4
Gene Name:
leucine rich repeat containing G protein-coupled receptor 4
Gene Aliases:
BNMD17, GPR48
Chromosome Location:
Chr.11: 27365961 - 27472787 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01551773_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LGR4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018490.2 NP_060960.2 14 - 15 1727 90
GenBank mRNA AB209743.1 - 14 - 15 1646 90
BC033039.2 - 13 - 14 1643 5260923 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100010400.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11001509.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene LGR4

Gene Ontology Categories:

Function(s) Process(es)

osteoblast differentiation
hair follicle development
G-protein coupled receptor signaling pathway
spermatogenesis
bone mineralization
male genitalia development
circadian regulation of gene expression
negative regulation of toll-like receptor signaling pathway
intestinal stem cell homeostasis
innate immune response
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
bone remodeling
digestive tract development
negative regulation of cytokine secretion
canonical Wnt signaling pathway involved in metanephric kidney development
cell differentiation involved in metanephros development
metanephric glomerulus development
metanephric nephron tubule morphogenesis
positive regulation of branching involved in ureteric bud morphogenesis
positive regulation of canonical Wnt signaling pathway
epithelial cell proliferation involved in renal tubule morphogenesis
transmembrane signaling receptor activity
G-protein coupled receptor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR11018888.hg.1 ENST00000379214.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11018888.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11018888.hg.1 ENST00000459658.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11018888.hg.1 NM_018490.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11018888.hg.1 TC11001509.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR11018889.hg.1 ENST00000379214.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11018889.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11018889.hg.1 ENST00000459658.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11018889.hg.1 NM_018490.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11018889.hg.1 TC11001509.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000480977.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000480977.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000480977.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000489910.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000489910.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000489910.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000527773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000527773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000527773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000527773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000530658.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000530658.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 ENST00000530658.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000480977.2 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000480977.2 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000480977.2 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000489910.1 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000489910.1 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000489910.1 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000527773.1 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000527773.1 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000527773.1 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000527773.1 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000530658.1 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000530658.1 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 ENST00000530658.1 Ensembl
Affymetrix Human Clariom D Assay TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Clariom D Assay TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Clariom D Assay TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Clariom D Assay TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Clariom D Assay TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Clariom D Assay TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Clariom D Assay TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Clariom D Assay TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Clariom D Assay TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Clariom D Assay TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Clariom D Assay TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Clariom D Assay TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Clariom D Assay TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Clariom D Assay TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Clariom D Assay TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Clariom D Assay TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Clariom D Assay TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Clariom D Assay TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Clariom D Assay TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Clariom D Assay TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Clariom D Assay TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Clariom D Assay TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Clariom D Assay TC1100010400.hg.1 NM_018490.3 Refseq
Affymetrix Human Clariom D Assay PSR1100178829.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100178829.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100178829.hg.1 ENST00000489910.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100178829.hg.1 NM_018490.3 Refseq
Affymetrix Human Clariom D Assay PSR1100178829.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100178829.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100178829.hg.1 ENST00000489910.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100178829.hg.1 NM_018490.3 Refseq
Affymetrix Human Clariom D Assay PSR1100178829.hg.1 TC1100010400.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100178830.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100178830.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100178830.hg.1 ENST00000489910.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100178830.hg.1 NM_018490.3 Refseq
Affymetrix Human Clariom D Assay PSR1100178830.hg.1 ENST00000379214.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100178830.hg.1 ENST00000389858.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100178830.hg.1 ENST00000489910.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100178830.hg.1 NM_018490.3 Refseq
Affymetrix Human Clariom D Assay PSR1100178830.hg.1 TC1100010400.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0