This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01550089_g1
See other PDCD1 GE Assays ›
Gene Symbol:
PDCD1
Gene Name:
programmed cell death 1
Gene Aliases:
CD279, PD-1, PD1, SLEB2, hPD-1, hPD-l, hSLE1
Chromosome Location:
Chr.2: 241849881 - 241858908 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01550089_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PDCD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005018.2 NP_005009.2 3 - 4 663 115
XM_017004293.1 XP_016859782.1 3 - 4 665 112
GenBank mRNA AK313848.1 - 3 - 4 665 115
AY238517.1 - 3 - 4 595 115
AY322430.1 - 385 115
BC074740.2 - 3 - 4 641 30915198 115
KJ865859.1 - 3 - 4 595 115
KJ865860.1 - 3 - 4 595 115
KJ865861.1 - 3 - 4 595 115
U64863.1 - 3 - 4 663 115

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200016382.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02002960.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PDCD1

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of tolerance induction
apoptotic process
humoral immune response
signal transduction
multicellular organism development
T cell costimulation
negative regulation of apoptotic process
positive regulation of T cell apoptotic process
signal transducer activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 ENST00000334409.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 ENST00000334409.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 ENST00000334409.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 ENST00000334409.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 ENST00000334409.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 ENST00000334409.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 ENST00000334409.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 ENST00000334409.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 ENST00000334409.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 ENST00000334409.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 ENST00000334409.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 ENST00000334409.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 ENST00000343705.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 ENST00000343705.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 ENST00000343705.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 ENST00000343705.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 ENST00000418831.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 ENST00000418831.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 ENST00000418831.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 ENST00000418831.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 ENST00000418831.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 ENST00000418831.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 ENST00000418831.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 ENST00000418831.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 ENST00000418831.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 ENST00000418831.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 NM_005018.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 NM_005018.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 NM_005018.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 NM_005018.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 NM_005018.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 NM_005018.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 NM_005018.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 NM_005018.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 NM_005018.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 NM_005018.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 NM_005018.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016382.hg.1 NM_005018.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0