This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01396860_m1
See other LOC101927432 GE Assays ›
Gene Symbol:
LOC101927432
Gene Name:
uncharacterized LOC101927432
Gene Aliases:
-
Chromosome Location:
Chr.3: 133333571 - 133386339 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01396860_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101927432
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC040919.1 - 5 - 6 648 84

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream TMEM108 antisense RNA 1 TMEM108-AS1 -107.8kb Sense Chr. 3 - 133246448 - 133257121 on NCBI Build GRCh38
Downstream beaded filament structural protein 2 BFSP2 CP47,
CP49,
CTRCT12,
LIFL-L,
PHAKOSIN
33.4kb Anti-sense Chr. 3 - 133397882 - 133475212 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0