This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01380112_m1
See other RIIAD1 GE Assays ›
Gene Symbol:
RIIAD1
Gene Name:
regulatory subunit of type II PKA R-subunit (RIIa) domain containing 1
Gene Aliases:
C1orf230, NCRNA00166
Chromosome Location:
Chr.1: 151721537 - 151729606 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01380112_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RIIAD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001144956.1 NP_001138428.1 3 - 4 108
GenBank mRNA AA393535.1 - 3 - 4 108
CK300240.1 - 2 - 3 267 108

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100009969.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01001217.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000326413.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000326413.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000326413.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000326413.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000326413.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000326413.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000326413.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000326413.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000326413.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000326413.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000326413.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000326413.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000426175.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000426175.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000426175.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000426175.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000426175.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000426175.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000427205.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000427205.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000427205.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000427205.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000427205.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000451222.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000451222.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000451222.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000451222.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000451222.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000451484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000451484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000451484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000451484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000451484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000451484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000451484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000451484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000479191.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000479191.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000479191.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000479191.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000479191.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000479191.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000479191.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 ENST00000479191.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 NM_001144956.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 NM_001144956.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 NM_001144956.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 NM_001144956.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 NM_001144956.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 NM_001144956.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 NM_001144956.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009969.hg.1 NM_001144956.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01018889.hg.1 ENST00000326413.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01018889.hg.1 ENST00000426175.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01018889.hg.1 ENST00000442233.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01018889.hg.1 ENST00000479191.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01018889.hg.1 NM_001144956.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01018889.hg.1 TC01001217.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000326413.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000326413.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000326413.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000326413.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000326413.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000326413.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000326413.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000326413.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000326413.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000326413.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000326413.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000326413.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000426175.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000426175.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000426175.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000426175.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000426175.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000426175.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000427205.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000427205.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000427205.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000427205.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000427205.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000451222.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000451222.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000451222.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000451222.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000451222.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000451484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000451484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000451484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000451484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000451484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000451484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000451484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000451484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000479191.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000479191.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000479191.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000479191.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000479191.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000479191.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000479191.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 ENST00000479191.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009969.hg.1 NM_001144956.1 Refseq
Affymetrix Human Clariom D Assay TC0100009969.hg.1 NM_001144956.1 Refseq
Affymetrix Human Clariom D Assay TC0100009969.hg.1 NM_001144956.1 Refseq
Affymetrix Human Clariom D Assay TC0100009969.hg.1 NM_001144956.1 Refseq
Affymetrix Human Clariom D Assay TC0100009969.hg.1 NM_001144956.1 Refseq
Affymetrix Human Clariom D Assay TC0100009969.hg.1 NM_001144956.1 Refseq
Affymetrix Human Clariom D Assay TC0100009969.hg.1 NM_001144956.1 Refseq
Affymetrix Human Clariom D Assay TC0100009969.hg.1 NM_001144956.1 Refseq
Affymetrix Human Clariom D Assay PSR0100172303.hg.1 ENST00000326413.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100172303.hg.1 ENST00000426175.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100172303.hg.1 ENST00000427205.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100172303.hg.1 ENST00000451222.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100172303.hg.1 ENST00000451484.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100172303.hg.1 ENST00000479191.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100172303.hg.1 NM_001144956.1 Refseq
Affymetrix Human Clariom D Assay PSR0100172303.hg.1 ENST00000326413.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100172303.hg.1 ENST00000426175.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100172303.hg.1 ENST00000427205.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100172303.hg.1 ENST00000451222.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100172303.hg.1 ENST00000451484.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100172303.hg.1 ENST00000479191.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100172303.hg.1 NM_001144956.1 Refseq
Affymetrix Human Clariom D Assay PSR0100172303.hg.1 TC0100009969.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0