This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01128444_m1
See other SLC45A2 GE Assays ›
Gene Symbol:
SLC45A2
Gene Name:
solute carrier family 45 member 2
Gene Aliases:
1A1, AIM1, MATP, OCA4, SHEP5
Chromosome Location:
Chr.5: 33944616 - 33984675 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01128444_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC45A2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001012509.3 NP_001012527.1 2 - 3 652 77
NM_016180.4 NP_057264.3 2 - 3 652 77
GenBank mRNA AF172849.1 - 2 - 3 585 77
BC064405.1 - 2 - 3 618 5563039 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500010456.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05001245.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLC45A2

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

visual perception
sucrose transport
developmental pigmentation
response to stimulus
sucrose:proton symporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000296589.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000296589.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000296589.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000296589.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000296589.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000296589.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000296589.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000296589.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000296589.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000296589.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000296589.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000296589.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000296589.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000296589.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000382102.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000382102.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000382102.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000382102.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000382102.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000382102.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000382102.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000382102.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000382102.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000382102.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000382102.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000505056.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000505056.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000505056.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000505056.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000509381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000509381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000509381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000509381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000509381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000509381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000509381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000509381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000510600.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000510600.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000510600.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000510600.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000510600.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 ENST00000510600.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 NM_001297417.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 NM_001297417.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 NM_001297417.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 NM_001297417.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 NM_001297417.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 NM_001297417.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 NM_001297417.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 NM_001297417.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500010456.hg.1 NM_001297417.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05017486.hg.1 ENST00000296589.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05017486.hg.1 ENST00000342059.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05017486.hg.1 ENST00000382102.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05017486.hg.1 NM_001012509.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05017486.hg.1 NM_016180.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05017486.hg.1 TC05001245.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0500172668.hg.1 ENST00000296589.8 Ensembl
Affymetrix Human Clariom D Assay PSR0500172668.hg.1 ENST00000382102.7 Ensembl
Affymetrix Human Clariom D Assay PSR0500172668.hg.1 ENST00000505056.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500172668.hg.1 ENST00000510600.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500172668.hg.1 ENST00000296589.8 Ensembl
Affymetrix Human Clariom D Assay PSR0500172668.hg.1 ENST00000382102.7 Ensembl
Affymetrix Human Clariom D Assay PSR0500172668.hg.1 ENST00000505056.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500172668.hg.1 ENST00000510600.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500172668.hg.1 TC0500010456.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000296589.8 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000296589.8 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000296589.8 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000296589.8 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000296589.8 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000296589.8 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000296589.8 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000296589.8 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000296589.8 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000296589.8 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000296589.8 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000296589.8 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000296589.8 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000296589.8 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000382102.7 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000382102.7 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000382102.7 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000382102.7 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000382102.7 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000382102.7 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000382102.7 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000382102.7 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000382102.7 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000382102.7 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000382102.7 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000505056.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000505056.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000505056.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000505056.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000509381.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000509381.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000509381.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000509381.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000509381.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000509381.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000509381.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000509381.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000510600.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000510600.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000510600.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000510600.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000510600.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 ENST00000510600.1 Ensembl
Affymetrix Human Clariom D Assay TC0500010456.hg.1 NM_001297417.2 Refseq
Affymetrix Human Clariom D Assay TC0500010456.hg.1 NM_001297417.2 Refseq
Affymetrix Human Clariom D Assay TC0500010456.hg.1 NM_001297417.2 Refseq
Affymetrix Human Clariom D Assay TC0500010456.hg.1 NM_001297417.2 Refseq
Affymetrix Human Clariom D Assay TC0500010456.hg.1 NM_001297417.2 Refseq
Affymetrix Human Clariom D Assay TC0500010456.hg.1 NM_001297417.2 Refseq
Affymetrix Human Clariom D Assay TC0500010456.hg.1 NM_001297417.2 Refseq
Affymetrix Human Clariom D Assay TC0500010456.hg.1 NM_001297417.2 Refseq
Affymetrix Human Clariom D Assay TC0500010456.hg.1 NM_001297417.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0