This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01127784_m1
See other PDE6C GE Assays ›
Gene Symbol:
PDE6C
Gene Name:
phosphodiesterase 6C
Gene Aliases:
ACHM5, COD4, PDEA2
Chromosome Location:
Chr.10: 93612588 - 93665673 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01127784_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PDE6C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_006204.3 NP_006195.3 7 - 8 1215 107
XM_017016322.1 XP_016871811.1 5 - 6 579 107

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000008488.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10000667.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PDE6C

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

visual perception
phototransduction, visible light
retinal cone cell development
cGMP binding
metal ion binding
3',5'-cyclic-GMP phosphodiesterase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR10007512.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10007512.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10007512.hg.1 TC10000667.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000475427.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000475427.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000475427.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 ENST00000475427.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay PSR1000157714.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay PSR1000157714.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay PSR1000157714.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay PSR1000157714.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay PSR1000157714.hg.1 TC1000008488.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000371447.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000475427.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000475427.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000475427.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 ENST00000475427.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq
Affymetrix Human Clariom D Assay TC1000008488.hg.1 NM_006204.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0