This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01116312_g1
See other PRPF38A GE Assays ›
Gene Symbol:
PRPF38A
Gene Name:
pre-mRNA processing factor 38A
Gene Aliases:
Prp38
Chromosome Location:
Chr.1: 52404547 - 52419723 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01116312_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRPF38A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK315621.1 - 1 - 2 314 92
BC000777.2 - 1 - 2 249 92
BC063655.1 - 1 - 2 306 92

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000474048.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000474048.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000474048.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000474048.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000474048.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000474048.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000474048.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000474048.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000474048.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000474048.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000474048.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000474048.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000487160.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000487160.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000487160.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000487160.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000487160.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000487160.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 ENST00000487160.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000257181.9 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000474048.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000474048.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000474048.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000474048.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000474048.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000474048.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000474048.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000474048.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000474048.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000474048.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000474048.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000474048.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000487160.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000487160.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000487160.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000487160.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000487160.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000487160.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 ENST00000487160.1 Ensembl
Affymetrix Human Clariom D Assay TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Clariom D Assay TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Clariom D Assay TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Clariom D Assay TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Clariom D Assay TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Clariom D Assay TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Clariom D Assay TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Clariom D Assay TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Clariom D Assay TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Clariom D Assay TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Clariom D Assay TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Clariom D Assay TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Clariom D Assay TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Clariom D Assay TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Clariom D Assay TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Clariom D Assay TC0100008315.hg.1 NM_032864.3 Refseq
Affymetrix Human Clariom D Assay TC0100008315.hg.1 NM_032864.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0