This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01115180_m1
See other PTPDC1 GE Assays ›
Gene Symbol:
PTPDC1
Gene Name:
protein tyrosine phosphatase domain containing 1
Gene Aliases:
PTP9Q22
Chromosome Location:
Chr.9: 94030591 - 94109856 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01115180_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PTPDC1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001253829.1 NP_001240758.1 4 - 5 715 122
NM_001253830.1 NP_001240759.1 5 - 6 776 122
NM_152422.4 NP_689635.3 4 - 5 709 122
NM_177995.2 NP_818931.1 5 - 6 791 122
XM_005251711.4 XP_005251768.1 5 - 6 2733 122
XM_011518240.2 XP_011516542.1 5 - 6 541 122
XM_011518241.2 XP_011516543.1 5 - 6 495 122
XM_017014280.1 XP_016869769.1 6 - 7 600 122
XM_017014281.1 XP_016869770.1 5 - 6 519 122
GenBank mRNA AK124912.1 - 4 - 5 595 122
AK289692.1 - 4 - 5 666 122
AK310403.1 - 4 - 5 673 122
AY171233.1 - 5 - 6 796 122
BC067120.1 - 5 - 6 776 5539182 122
DA312531.1 - 3 - 4 291 122

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900008010.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09000455.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PTPDC1

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

peptidyl-tyrosine dephosphorylation
cilium morphogenesis
protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR09004900.hg.1 ENST00000288976.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09004900.hg.1 ENST00000375360.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09004900.hg.1 NM_001253829.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09004900.hg.1 NM_001253830.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09004900.hg.1 NM_152422.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09004900.hg.1 NM_177995.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09004900.hg.1 TC09000455.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000288976.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000288976.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000288976.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000288976.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000288976.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000288976.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000288976.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000288976.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000288976.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000288976.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000288976.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000288976.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000375360.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000375360.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000375360.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000375360.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000375360.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000375360.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000375360.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000375360.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000375360.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000375360.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000375360.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000375360.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000467049.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000467049.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000467049.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000467049.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000620992.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000620992.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000620992.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000620992.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000620992.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000620992.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000620992.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000620992.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000620992.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000620992.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000620992.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000620992.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 ENST00000620992.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_001253829.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_001253829.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_001253829.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_001253829.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_001253829.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_001253829.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_001253829.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_001253829.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_001253829.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_001253829.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_001253829.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_001253829.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_001253829.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_001253830.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_001253830.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_001253830.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_001253830.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_001253830.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_001253830.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_001253830.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_001253830.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_001253830.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_001253830.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_001253830.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_001253830.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_152422.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_152422.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_152422.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_152422.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_152422.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_152422.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_152422.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_152422.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_152422.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_152422.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_152422.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_152422.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_152422.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_177995.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_177995.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_177995.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_177995.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_177995.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_177995.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_177995.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_177995.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_177995.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_177995.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_177995.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008010.hg.1 NM_177995.2 Refseq
Affymetrix Human Clariom D Assay PSR0900153942.hg.1 ENST00000288976.3 Ensembl
Affymetrix Human Clariom D Assay PSR0900153942.hg.1 ENST00000375360.7 Ensembl
Affymetrix Human Clariom D Assay PSR0900153942.hg.1 ENST00000620992.4 Ensembl
Affymetrix Human Clariom D Assay PSR0900153942.hg.1 NM_001253829.1 Refseq
Affymetrix Human Clariom D Assay PSR0900153942.hg.1 NM_001253830.1 Refseq
Affymetrix Human Clariom D Assay PSR0900153942.hg.1 NM_152422.4 Refseq
Affymetrix Human Clariom D Assay PSR0900153942.hg.1 NM_177995.2 Refseq
Affymetrix Human Clariom D Assay PSR0900153942.hg.1 ENST00000288976.3 Ensembl
Affymetrix Human Clariom D Assay PSR0900153942.hg.1 ENST00000375360.7 Ensembl
Affymetrix Human Clariom D Assay PSR0900153942.hg.1 ENST00000620992.4 Ensembl
Affymetrix Human Clariom D Assay PSR0900153942.hg.1 NM_001253829.1 Refseq
Affymetrix Human Clariom D Assay PSR0900153942.hg.1 NM_001253830.1 Refseq
Affymetrix Human Clariom D Assay PSR0900153942.hg.1 NM_152422.4 Refseq
Affymetrix Human Clariom D Assay PSR0900153942.hg.1 NM_177995.2 Refseq
Affymetrix Human Clariom D Assay PSR0900153942.hg.1 TC0900008010.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000288976.3 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000288976.3 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000288976.3 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000288976.3 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000288976.3 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000288976.3 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000288976.3 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000288976.3 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000288976.3 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000288976.3 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000288976.3 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000288976.3 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000375360.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000375360.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000375360.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000375360.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000375360.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000375360.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000375360.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000375360.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000375360.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000375360.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000375360.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000375360.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000467049.1 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000467049.1 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000467049.1 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000467049.1 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000620992.4 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000620992.4 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000620992.4 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000620992.4 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000620992.4 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000620992.4 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000620992.4 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000620992.4 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000620992.4 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000620992.4 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000620992.4 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000620992.4 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 ENST00000620992.4 Ensembl
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_001253829.1 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_001253829.1 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_001253829.1 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_001253829.1 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_001253829.1 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_001253829.1 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_001253829.1 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_001253829.1 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_001253829.1 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_001253829.1 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_001253829.1 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_001253829.1 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_001253829.1 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_001253830.1 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_001253830.1 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_001253830.1 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_001253830.1 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_001253830.1 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_001253830.1 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_001253830.1 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_001253830.1 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_001253830.1 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_001253830.1 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_001253830.1 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_001253830.1 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_152422.4 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_152422.4 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_152422.4 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_152422.4 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_152422.4 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_152422.4 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_152422.4 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_152422.4 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_152422.4 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_152422.4 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_152422.4 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_152422.4 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_152422.4 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_177995.2 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_177995.2 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_177995.2 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_177995.2 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_177995.2 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_177995.2 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_177995.2 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_177995.2 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_177995.2 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_177995.2 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_177995.2 Refseq
Affymetrix Human Clariom D Assay TC0900008010.hg.1 NM_177995.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0