This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01111213_s1
See other AMER2 GE Assays ›
Gene Symbol:
AMER2
Gene Name:
APC membrane recruitment protein 2
Gene Aliases:
FAM123A
Chromosome Location:
Chr.13: 25161679 - 25172456 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01111213_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AMER2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_152704.3 NP_689917.2 1 5021 87
NM_199138.2 NP_954589.1 3 4355 87
XM_005266279.3 XP_005266336.1 2 5001 87
XM_005266280.3 XP_005266337.1 2 4911 87
XM_017020415.1 XP_016875904.1 2 4675 87
GenBank mRNA AL832586.1 - 1 436 87
AL832627.1 - 1 3075 87

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300008381.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene AMER2

Gene Ontology Categories:

Function(s) Process(es)

ectoderm development
Wnt signaling pathway
anatomical structure development
negative regulation of canonical Wnt signaling pathway
protein binding
phosphatidylinositol-4,5-bisphosphate binding
beta-catenin binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 ENST00000357816.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 ENST00000357816.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 ENST00000357816.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 ENST00000357816.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 ENST00000357816.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 ENST00000357816.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 ENST00000357816.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 ENST00000357816.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 ENST00000515384.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 ENST00000515384.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 ENST00000515384.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 ENST00000515384.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 ENST00000515384.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 ENST00000515384.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 ENST00000515384.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 ENST00000515384.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 NM_152704.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 NM_152704.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 NM_152704.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 NM_152704.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 NM_152704.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 NM_152704.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 NM_152704.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 NM_152704.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 NM_152704.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 NM_152704.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 NM_199138.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 NM_199138.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 NM_199138.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 NM_199138.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 NM_199138.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 NM_199138.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008381.hg.1 NM_199138.2 Refseq
Affymetrix Human Clariom D Assay PSR1300156572.hg.1 ENST00000357816.2 Ensembl
Affymetrix Human Clariom D Assay PSR1300156572.hg.1 NM_152704.3 Refseq
Affymetrix Human Clariom D Assay PSR1300156572.hg.1 NM_199138.2 Refseq
Affymetrix Human Clariom D Assay PSR1300156572.hg.1 ENST00000357816.2 Ensembl
Affymetrix Human Clariom D Assay PSR1300156572.hg.1 NM_152704.3 Refseq
Affymetrix Human Clariom D Assay PSR1300156572.hg.1 NM_199138.2 Refseq
Affymetrix Human Clariom D Assay PSR1300156572.hg.1 TC1300008381.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1300008381.hg.1 ENST00000357816.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008381.hg.1 ENST00000357816.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008381.hg.1 ENST00000357816.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008381.hg.1 ENST00000357816.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008381.hg.1 ENST00000357816.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008381.hg.1 ENST00000357816.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008381.hg.1 ENST00000357816.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008381.hg.1 ENST00000357816.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008381.hg.1 ENST00000515384.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008381.hg.1 ENST00000515384.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008381.hg.1 ENST00000515384.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008381.hg.1 ENST00000515384.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008381.hg.1 ENST00000515384.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008381.hg.1 ENST00000515384.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008381.hg.1 ENST00000515384.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008381.hg.1 ENST00000515384.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008381.hg.1 NM_152704.3 Refseq
Affymetrix Human Clariom D Assay TC1300008381.hg.1 NM_152704.3 Refseq
Affymetrix Human Clariom D Assay TC1300008381.hg.1 NM_152704.3 Refseq
Affymetrix Human Clariom D Assay TC1300008381.hg.1 NM_152704.3 Refseq
Affymetrix Human Clariom D Assay TC1300008381.hg.1 NM_152704.3 Refseq
Affymetrix Human Clariom D Assay TC1300008381.hg.1 NM_152704.3 Refseq
Affymetrix Human Clariom D Assay TC1300008381.hg.1 NM_152704.3 Refseq
Affymetrix Human Clariom D Assay TC1300008381.hg.1 NM_152704.3 Refseq
Affymetrix Human Clariom D Assay TC1300008381.hg.1 NM_152704.3 Refseq
Affymetrix Human Clariom D Assay TC1300008381.hg.1 NM_152704.3 Refseq
Affymetrix Human Clariom D Assay TC1300008381.hg.1 NM_199138.2 Refseq
Affymetrix Human Clariom D Assay TC1300008381.hg.1 NM_199138.2 Refseq
Affymetrix Human Clariom D Assay TC1300008381.hg.1 NM_199138.2 Refseq
Affymetrix Human Clariom D Assay TC1300008381.hg.1 NM_199138.2 Refseq
Affymetrix Human Clariom D Assay TC1300008381.hg.1 NM_199138.2 Refseq
Affymetrix Human Clariom D Assay TC1300008381.hg.1 NM_199138.2 Refseq
Affymetrix Human Clariom D Assay TC1300008381.hg.1 NM_199138.2 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 232021 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header