This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01110003_s1
See other BACH1 GE Assays ›
Gene Symbol:
BACH1
Gene Name:
BTB domain and CNC homolog 1
Gene Aliases:
BACH-1, BTBD24
Chromosome Location:
Chr.21: 29298795 - 29361896 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01110003_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BACH1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_027655.2 5 2004 63
GenBank mRNA AF317902.1 - 1 49 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2100008490.hg.1
This assay targets the same location as HTA transcript cluster(s) TC21000097.hg.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription involved in G1/S transition of mitotic cell cycle
regulation of transcription involved in G2/M transition of mitotic cell cycle
negative regulation of transcription from RNA polymerase II promoter
DNA repair
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
protein ubiquitination
positive regulation of transcription from RNA polymerase II promoter
regulation of transcription from RNA polymerase II promoter in response to hypoxia
RNA polymerase II distal enhancer sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
transcriptional repressor activity, RNA polymerase II distal enhancer sequence-specific binding
transcription factor activity, sequence-specific DNA binding
ubiquitin-protein transferase activity
protein binding
heme binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR21000617.hg.1 ENST00000504298.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR21000617.hg.1 NR_027655.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR21000617.hg.1 TC21000097.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR21000619.hg.1 ENST00000504298.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR21000619.hg.1 NR_027655.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR21000619.hg.1 TC21000097.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR21000616.hg.1 ENST00000504298.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR21000616.hg.1 NR_027655.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR21000616.hg.1 TC21000097.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR21000618.hg.1 ENST00000504298.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR21000618.hg.1 NR_027655.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR21000618.hg.1 TC21000097.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000286800.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000286800.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000286800.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000286800.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000286800.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000286800.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000286800.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000286800.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000286800.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000286800.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000286800.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000286800.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000399921.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000399921.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000399921.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000399921.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000399921.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000399921.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000399921.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000399921.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000399921.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000399921.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000399921.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000435072.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000435072.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000435072.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000435072.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000435072.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000435072.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000447177.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000447177.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000447177.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000447177.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000447177.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000447177.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000451655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000451655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000451655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000451655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000451655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000451655.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000504298.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000504298.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000504298.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000504298.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000504298.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000548467.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000548467.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000548467.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 ENST00000609910.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NM_001186.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NM_001186.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NM_001186.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NM_001186.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NM_001186.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NM_001186.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NM_001186.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NM_001186.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NM_001186.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NM_001186.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NM_001186.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NM_001186.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NM_001186.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NM_206866.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NM_206866.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NM_206866.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NM_206866.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NM_206866.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NM_206866.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NM_206866.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NM_206866.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NM_206866.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NM_206866.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NM_206866.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NR_027655.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NR_027655.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NR_027655.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NR_027655.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NR_027655.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NR_027655.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NR_027655.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NR_027655.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NR_027655.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NR_027655.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NR_027655.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NR_027655.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NR_027655.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008490.hg.1 NR_027655.2 Refseq
Affymetrix Human Clariom D Assay PSR2100157823.hg.1 ENST00000504298.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100157823.hg.1 NR_027655.2 Refseq
Affymetrix Human Clariom D Assay PSR2100157823.hg.1 ENST00000504298.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100157823.hg.1 NR_027655.2 Refseq
Affymetrix Human Clariom D Assay PSR2100157823.hg.1 TC2100008490.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000286800.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000286800.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000286800.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000286800.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000286800.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000286800.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000286800.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000286800.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000286800.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000286800.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000286800.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000286800.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000399921.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000399921.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000399921.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000399921.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000399921.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000399921.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000399921.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000399921.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000399921.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000399921.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000399921.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000435072.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000435072.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000435072.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000435072.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000435072.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000435072.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000447177.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000447177.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000447177.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000447177.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000447177.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000447177.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000451655.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000451655.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000451655.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000451655.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000451655.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000451655.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000504298.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000504298.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000504298.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000504298.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000504298.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000548467.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000548467.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000548467.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 ENST00000609910.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NM_001186.3 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NM_001186.3 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NM_001186.3 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NM_001186.3 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NM_001186.3 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NM_001186.3 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NM_001186.3 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NM_001186.3 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NM_001186.3 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NM_001186.3 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NM_001186.3 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NM_001186.3 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NM_001186.3 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NM_206866.2 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NM_206866.2 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NM_206866.2 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NM_206866.2 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NM_206866.2 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NM_206866.2 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NM_206866.2 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NM_206866.2 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NM_206866.2 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NM_206866.2 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NM_206866.2 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NR_027655.2 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NR_027655.2 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NR_027655.2 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NR_027655.2 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NR_027655.2 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NR_027655.2 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NR_027655.2 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NR_027655.2 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NR_027655.2 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NR_027655.2 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NR_027655.2 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NR_027655.2 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NR_027655.2 Refseq
Affymetrix Human Clariom D Assay TC2100008490.hg.1 NR_027655.2 Refseq
Affymetrix Human Clariom D Assay PSR2100157831.hg.1 ENST00000504298.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100157831.hg.1 NR_027655.2 Refseq
Affymetrix Human Clariom D Assay PSR2100157831.hg.1 ENST00000504298.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100157831.hg.1 NR_027655.2 Refseq
Affymetrix Human Clariom D Assay PSR2100157831.hg.1 TC2100008490.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2100157821.hg.1 ENST00000504298.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100157821.hg.1 NR_027655.2 Refseq
Affymetrix Human Clariom D Assay PSR2100157821.hg.1 ENST00000504298.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100157821.hg.1 NR_027655.2 Refseq
Affymetrix Human Clariom D Assay PSR2100157821.hg.1 TC2100008490.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2100157827.hg.1 ENST00000504298.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100157827.hg.1 NR_027655.2 Refseq
Affymetrix Human Clariom D Assay PSR2100157827.hg.1 ENST00000504298.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100157827.hg.1 NR_027655.2 Refseq
Affymetrix Human Clariom D Assay PSR2100157827.hg.1 TC2100008490.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2100157829.hg.1 ENST00000504298.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100157829.hg.1 NR_027655.2 Refseq
Affymetrix Human Clariom D Assay PSR2100157829.hg.1 ENST00000504298.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100157829.hg.1 NR_027655.2 Refseq
Affymetrix Human Clariom D Assay PSR2100157829.hg.1 TC2100008490.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 575026 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0