This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01103667_m1
See other MRPL1 GE Assays ›
Gene Symbol:
MRPL1
Gene Name:
mitochondrial ribosomal protein L1
Gene Aliases:
BM022, L1MT, MRP-L1
Chromosome Location:
Chr.4: 77861955 - 77952790 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01103667_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MRPL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_020236.3 NP_064621.3 5 - 6 760 67
XM_011532198.2 XP_011530500.1 6 - 7 812 67
XM_011532199.1 XP_011530501.1 6 - 7 626 67
GenBank mRNA AB049474.1 - 4 - 5 534 67
AF212225.1 - 5 - 6 664 67
AK024791.1 - 3 - 4 363 67
AK315600.1 - 5 - 6 577 67
BC014356.1 - 5 - 6 584 3686618 67
BC015109.1 - 5 - 6 573 4044993 67
BC017765.1 - 5 - 6 581 67
BC032595.1 - 5 - 6 571 5527878 67
BU675164.1 - 4 - 5 67
CR457191.1 - 4 - 5 496 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400007923.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04000433.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MRPL1

Panther Classification:

Molecular Function -

ribosomal protein translational protein

Gene Ontology Categories:

Function(s) Process(es)

maturation of LSU-rRNA
translation
mitochondrial translational elongation
mitochondrial translational termination
RNA binding
structural constituent of ribosome
protein binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR04006148.hg.1 ENST00000315567.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04006148.hg.1 ENST00000506674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04006148.hg.1 ENST00000538314.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04006148.hg.1 NM_020236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04006148.hg.1 TC04000433.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000315567.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000315567.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000315567.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000315567.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000315567.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000315567.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000315567.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000315567.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000315567.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000315567.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000315567.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000315567.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000315567.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000502384.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000502384.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000502384.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000502384.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000502384.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000502384.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000502384.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000502384.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000504901.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000504901.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000504901.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000504901.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000504901.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000504901.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000504901.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000506674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000506674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000506674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000506674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000506674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000506674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000511521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000511521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000511521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000511521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 ENST00000515625.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 NM_020236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 NM_020236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 NM_020236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 NM_020236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 NM_020236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 NM_020236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 NM_020236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 NM_020236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 NM_020236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 NM_020236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 NM_020236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007923.hg.1 NM_020236.3 Refseq
Affymetrix Human Clariom D Assay PSR0400154419.hg.1 ENST00000315567.12 Ensembl
Affymetrix Human Clariom D Assay PSR0400154419.hg.1 ENST00000502384.3 Ensembl
Affymetrix Human Clariom D Assay PSR0400154419.hg.1 ENST00000506674.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400154419.hg.1 NM_020236.3 Refseq
Affymetrix Human Clariom D Assay PSR0400154419.hg.1 ENST00000315567.12 Ensembl
Affymetrix Human Clariom D Assay PSR0400154419.hg.1 ENST00000502384.3 Ensembl
Affymetrix Human Clariom D Assay PSR0400154419.hg.1 ENST00000506674.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400154419.hg.1 NM_020236.3 Refseq
Affymetrix Human Clariom D Assay PSR0400154419.hg.1 TC0400007923.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000315567.12 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000315567.12 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000315567.12 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000315567.12 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000315567.12 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000315567.12 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000315567.12 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000315567.12 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000315567.12 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000315567.12 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000315567.12 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000315567.12 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000315567.12 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000502384.3 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000502384.3 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000502384.3 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000502384.3 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000502384.3 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000502384.3 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000502384.3 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000502384.3 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000504901.2 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000504901.2 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000504901.2 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000504901.2 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000504901.2 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000504901.2 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000504901.2 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000506674.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000506674.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000506674.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000506674.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000506674.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000506674.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000511521.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000511521.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000511521.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000511521.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 ENST00000515625.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007923.hg.1 NM_020236.3 Refseq
Affymetrix Human Clariom D Assay TC0400007923.hg.1 NM_020236.3 Refseq
Affymetrix Human Clariom D Assay TC0400007923.hg.1 NM_020236.3 Refseq
Affymetrix Human Clariom D Assay TC0400007923.hg.1 NM_020236.3 Refseq
Affymetrix Human Clariom D Assay TC0400007923.hg.1 NM_020236.3 Refseq
Affymetrix Human Clariom D Assay TC0400007923.hg.1 NM_020236.3 Refseq
Affymetrix Human Clariom D Assay TC0400007923.hg.1 NM_020236.3 Refseq
Affymetrix Human Clariom D Assay TC0400007923.hg.1 NM_020236.3 Refseq
Affymetrix Human Clariom D Assay TC0400007923.hg.1 NM_020236.3 Refseq
Affymetrix Human Clariom D Assay TC0400007923.hg.1 NM_020236.3 Refseq
Affymetrix Human Clariom D Assay TC0400007923.hg.1 NM_020236.3 Refseq
Affymetrix Human Clariom D Assay TC0400007923.hg.1 NM_020236.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0