This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01096668_m1
See other TGM2 GE Assays ›
Gene Symbol:
TGM2
Gene Name:
transglutaminase 2
Gene Aliases:
TG(C), TGC
Chromosome Location:
Chr.20: 38127387 - 38166578 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01096668_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TGM2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001323316.1 NP_001310245.1 11 - 12 1832 77
NM_001323317.1 NP_001310246.1 9 - 10 1606 77
NM_001323318.1 NP_001310247.1 9 - 10 1669 77
NM_004613.3 NP_004604.2 10 - 11 1849 77
XM_011529028.1 XP_011527330.1 11 - 12 1794 77
GenBank mRNA AK058031.1 - 10 - 11 1617 77
AK291714.1 - 10 - 11 1690 77
AK295775.1 - 9 - 10 1447 77
AK300292.1 - 9 - 10 1510 77
AK314618.1 - 10 - 11 1690 77
AY675221.1 - 9 - 10 1618 77
BQ938957.1 - 2 - 3 410 77
EU794665.1 - 10 - 11 1718 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2000009058.hg.1
This assay targets the same location as HTA transcript cluster(s) TC20000833.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TGM2

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

phospholipase C-activating G-protein coupled receptor signaling pathway
peptide cross-linking
negative regulation of endoplasmic reticulum calcium ion concentration
positive regulation of apoptotic process
apoptotic cell clearance
positive regulation of cell adhesion
positive regulation of mitochondrial calcium ion concentration
branching involved in salivary gland morphogenesis
salivary gland cavitation
protein-glutamine gamma-glutamyltransferase activity
protein binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR20011454.hg.1 ENST00000361475.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20011454.hg.1 ENST00000536701.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20011454.hg.1 ENST00000536724.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20011454.hg.1 NM_004613.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20011454.hg.1 TC20000833.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000373403.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000373403.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000373403.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000373403.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000373403.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000373403.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000373403.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000373403.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000373403.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000453095.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000453095.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000453095.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000453095.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000453095.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000453095.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000468262.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000468262.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000468262.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000468262.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000468262.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000468262.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000468262.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000468262.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000468262.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000468262.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000468262.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000468262.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000468262.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000469269.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000469269.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000469269.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000469269.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000469269.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000474777.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000474777.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000474777.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000474777.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000474777.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000474777.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000474777.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 ENST00000474777.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 NM_004613.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 NM_004613.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 NM_004613.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 NM_004613.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 NM_004613.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 NM_004613.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 NM_004613.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 NM_004613.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 NM_004613.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 NM_004613.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 NM_004613.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 NM_004613.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 NM_004613.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 NM_004613.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 NM_004613.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 NM_198951.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 NM_198951.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 NM_198951.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 NM_198951.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 NM_198951.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 NM_198951.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 NM_198951.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 NM_198951.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 NM_198951.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 NM_198951.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 NM_198951.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 NM_198951.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009058.hg.1 NM_198951.2 Refseq
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000373403.7 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000373403.7 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000373403.7 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000373403.7 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000373403.7 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000373403.7 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000373403.7 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000373403.7 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000373403.7 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000453095.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000453095.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000453095.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000453095.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000453095.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000453095.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000468262.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000468262.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000468262.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000468262.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000468262.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000468262.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000468262.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000468262.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000468262.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000468262.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000468262.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000468262.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000468262.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000469269.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000469269.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000469269.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000469269.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000469269.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000474777.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000474777.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000474777.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000474777.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000474777.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000474777.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000474777.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 ENST00000474777.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009058.hg.1 NM_004613.3 Refseq
Affymetrix Human Clariom D Assay TC2000009058.hg.1 NM_004613.3 Refseq
Affymetrix Human Clariom D Assay TC2000009058.hg.1 NM_004613.3 Refseq
Affymetrix Human Clariom D Assay TC2000009058.hg.1 NM_004613.3 Refseq
Affymetrix Human Clariom D Assay TC2000009058.hg.1 NM_004613.3 Refseq
Affymetrix Human Clariom D Assay TC2000009058.hg.1 NM_004613.3 Refseq
Affymetrix Human Clariom D Assay TC2000009058.hg.1 NM_004613.3 Refseq
Affymetrix Human Clariom D Assay TC2000009058.hg.1 NM_004613.3 Refseq
Affymetrix Human Clariom D Assay TC2000009058.hg.1 NM_004613.3 Refseq
Affymetrix Human Clariom D Assay TC2000009058.hg.1 NM_004613.3 Refseq
Affymetrix Human Clariom D Assay TC2000009058.hg.1 NM_004613.3 Refseq
Affymetrix Human Clariom D Assay TC2000009058.hg.1 NM_004613.3 Refseq
Affymetrix Human Clariom D Assay TC2000009058.hg.1 NM_004613.3 Refseq
Affymetrix Human Clariom D Assay TC2000009058.hg.1 NM_004613.3 Refseq
Affymetrix Human Clariom D Assay TC2000009058.hg.1 NM_004613.3 Refseq
Affymetrix Human Clariom D Assay TC2000009058.hg.1 NM_198951.2 Refseq
Affymetrix Human Clariom D Assay TC2000009058.hg.1 NM_198951.2 Refseq
Affymetrix Human Clariom D Assay TC2000009058.hg.1 NM_198951.2 Refseq
Affymetrix Human Clariom D Assay TC2000009058.hg.1 NM_198951.2 Refseq
Affymetrix Human Clariom D Assay TC2000009058.hg.1 NM_198951.2 Refseq
Affymetrix Human Clariom D Assay TC2000009058.hg.1 NM_198951.2 Refseq
Affymetrix Human Clariom D Assay TC2000009058.hg.1 NM_198951.2 Refseq
Affymetrix Human Clariom D Assay TC2000009058.hg.1 NM_198951.2 Refseq
Affymetrix Human Clariom D Assay TC2000009058.hg.1 NM_198951.2 Refseq
Affymetrix Human Clariom D Assay TC2000009058.hg.1 NM_198951.2 Refseq
Affymetrix Human Clariom D Assay TC2000009058.hg.1 NM_198951.2 Refseq
Affymetrix Human Clariom D Assay TC2000009058.hg.1 NM_198951.2 Refseq
Affymetrix Human Clariom D Assay TC2000009058.hg.1 NM_198951.2 Refseq
Affymetrix Human Clariom D Assay PSR2000163483.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Clariom D Assay PSR2000163483.hg.1 ENST00000469269.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000163483.hg.1 NM_004613.3 Refseq
Affymetrix Human Clariom D Assay PSR2000163483.hg.1 ENST00000361475.6 Ensembl
Affymetrix Human Clariom D Assay PSR2000163483.hg.1 ENST00000469269.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000163483.hg.1 NM_004613.3 Refseq
Affymetrix Human Clariom D Assay PSR2000163483.hg.1 TC2000009058.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0