This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01096627_m1
See other CDH22 GE Assays ›
Gene Symbol:
CDH22
Gene Name:
cadherin 22
Gene Aliases:
C20orf25, dJ998H6.1
Chromosome Location:
Chr.20: 46173733 - 46308498 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01096627_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDH22
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021248.2 NP_067071.1 10 - 11 2307 85
XM_011528994.2 XP_011527296.1 10 - 11 2266 85
XM_011528995.1 XP_011527297.1 10 - 11 2071 85
GenBank mRNA AF035300.1 - 3 - 4 322 85
AK289584.1 - 10 - 11 2307 85
BC039056.1 - 2 - 3 252 85
BC136526.1 - 9 - 10 2056 9020516 85

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2000009263.hg.1
This assay targets the same location as HTA transcript cluster(s) TC20000898.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CDH22

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

homophilic cell adhesion via plasma membrane adhesion molecules
brain development
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR20012266.hg.1 ENST00000372262.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20012266.hg.1 ENST00000537909.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20012266.hg.1 NM_021248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20012266.hg.1 TC20000898.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000372262.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000372262.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000372262.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000372262.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000372262.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000372262.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000372262.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000372262.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000372262.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000372262.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000372262.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000372262.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000372262.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000372262.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000474438.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000474438.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000474438.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000474438.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000474438.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000474438.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 NM_021248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 NM_021248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 NM_021248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 NM_021248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 NM_021248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 NM_021248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 NM_021248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 NM_021248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 NM_021248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 NM_021248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 NM_021248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 NM_021248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 NM_021248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 NM_021248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009263.hg.1 NM_021248.2 Refseq
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000372262.7 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000372262.7 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000372262.7 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000372262.7 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000372262.7 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000372262.7 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000372262.7 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000372262.7 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000372262.7 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000372262.7 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000372262.7 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000372262.7 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000372262.7 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000372262.7 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000474438.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000474438.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000474438.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000474438.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000474438.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000474438.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Clariom D Assay TC2000009263.hg.1 NM_021248.2 Refseq
Affymetrix Human Clariom D Assay TC2000009263.hg.1 NM_021248.2 Refseq
Affymetrix Human Clariom D Assay TC2000009263.hg.1 NM_021248.2 Refseq
Affymetrix Human Clariom D Assay TC2000009263.hg.1 NM_021248.2 Refseq
Affymetrix Human Clariom D Assay TC2000009263.hg.1 NM_021248.2 Refseq
Affymetrix Human Clariom D Assay TC2000009263.hg.1 NM_021248.2 Refseq
Affymetrix Human Clariom D Assay TC2000009263.hg.1 NM_021248.2 Refseq
Affymetrix Human Clariom D Assay TC2000009263.hg.1 NM_021248.2 Refseq
Affymetrix Human Clariom D Assay TC2000009263.hg.1 NM_021248.2 Refseq
Affymetrix Human Clariom D Assay TC2000009263.hg.1 NM_021248.2 Refseq
Affymetrix Human Clariom D Assay TC2000009263.hg.1 NM_021248.2 Refseq
Affymetrix Human Clariom D Assay TC2000009263.hg.1 NM_021248.2 Refseq
Affymetrix Human Clariom D Assay TC2000009263.hg.1 NM_021248.2 Refseq
Affymetrix Human Clariom D Assay TC2000009263.hg.1 NM_021248.2 Refseq
Affymetrix Human Clariom D Assay TC2000009263.hg.1 NM_021248.2 Refseq
Affymetrix Human Clariom D Assay PSR2000164845.hg.1 ENST00000372262.7 Ensembl
Affymetrix Human Clariom D Assay PSR2000164845.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Clariom D Assay PSR2000164845.hg.1 NM_021248.2 Refseq
Affymetrix Human Clariom D Assay PSR2000164845.hg.1 ENST00000372262.7 Ensembl
Affymetrix Human Clariom D Assay PSR2000164845.hg.1 ENST00000537909.2 Ensembl
Affymetrix Human Clariom D Assay PSR2000164845.hg.1 NM_021248.2 Refseq
Affymetrix Human Clariom D Assay PSR2000164845.hg.1 TC2000009263.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0